Orders LinkContactUsLinkComponent
Salt meat broth
Salt meat broth
Salt meat broth, 500 g
Katalog # HIMEM155-500G
Leverandør:   HIMEDIA
Salt meat broth
Katalog # HIMEM155-500G
Leverandør:  HIMEDIA
CAS nummer:  
Salt meat broth
Salt meat broth, 500 g
CAS nummer:  
Price
877,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
An enrichment medium for the isolation of Staphylococci from grossly contaminated specimens.
  • Can detect even low numbers of Staphylococci from samples
  • Sodium chloride maintains osmotic equilibrium
  • Peptic digest of animal tissue and beef extract provide essential nutrients
The medium is sensitive as it can detect even low numbers of Staphylococci from samples having large proportions of heterogeneous microbial flora. Peptic digest of animal tissue and beef extract provide essential nutrients for bacterial metabolism. Sodium chloride maintains osmotic equilibrium.
Specifikationer
Specifikationer
  • Pakn:
    500 g
  • Beskrivelse:
    Salt meat broth
  • Cat. No.:
    HIMEM155-500G
  • pH:
    7,4 - 7,8
Specifikationer