Rack for serological pipettes
Rack for serological pipettes (1 - 100 ml), klar
WHEA72190-0000
Manufactured from heavy-duty clear acrylic. Ideal for the storage of Wheaton serological pipettes from 1 to 100 ml.
Comprises five angled compartments. Top 3 compartments are 3" x 3" and bottom 2 compartments are 3" wide x 1.3/8" high. Also accommodates two pipette controllers on the top - diameter of hole for these is 7/8".
- Farve:klar
- Beskrivelse:Rack for serological pipettes (1 - 100 ml)