Acetanilide Melting Point Standard, Reference Standard
Acetanilide Melting Point Standard, ~114 °C, USP reference standard, Vial, 1 g
Katalog # 1004001.
Leverandør: USP
Acetanilide Melting Point Standard, Reference Standard
Katalog # 1004001.
Leverandør: USP
CAS nummer:
Gå til:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifikationer
Specifikationer
- Pakning:Vial
- Pakn:1 g
- Standard:USP reference standard
- Beskrivelse:Acetanilide Melting Point Standard
- Smeltepunkt:~114 °C
- Cat. No.:1004001.
- Beskrivelse:Acetanilide
- CAS-nr:103-84-4