Orders LinkContactUsLinkComponent
Sulbactam Related Compounds, Reference Standard
Sulbactam Related Compounds, Reference Standard
Supplier:   USP
Sulbactam Related Compounds, Reference Standard
Supplier:  USP
Certificates
Sulbactam Related Compounds, Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1623749.
  • 1623772.
  • 1623783.
  • 1623794.
  • Beskrivelse:
  • Sulbactam Related Compound A
  • Sulbactam Related Compound D
  • Sulbactam Related Compound E
  • Sulbactam Related Compound F
  • Synonym(s):
  • (2S)-2-Amino-3-methyl-3-sulFood Ingredientsnobutanoic acid
  • Potassium (2S,5R,6R)-6-bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
  • (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide
  • (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
  • CAS-nr:
  • 23315-18-6
  • 4027-64-9
  • 76646-91-8
  • 24158-88-1
  • Formel:
  • C₅H₁₁NO₄S
  • C₈H₉BrKNO₃S
  • C₈H₉Br₂NO₅S
  • C₈H₉Br₂NO₃S
Specifications
Product Family Options
Product Information
  • Beskrivelse
    Standard
    Pakning
    Pakn
    Availability
    Price
  • Sulbactam Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 10.020,00 kr
    10.020,00 kr
    1 * 15 mg
       10.020,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    1623749.
    Beskrivelse
    Sulbactam Related Compound A
    Synonym(s)
    (2S)-2-Amino-3-methyl-3-sulFood Ingredientsnobutanoic acid
    CAS-nr
    23315-18-6
    Formel
    C₅H₁₁NO₄S
  • Sulbactam Related Compound D
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 10.520,00 kr
    10.520,00 kr
    1 * 15 mg
       10.520,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    1623772.
    Beskrivelse
    Sulbactam Related Compound D
    Synonym(s)
    Potassium (2S,5R,6R)-6-bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
    CAS-nr
    4027-64-9
    Formel
    C₈H₉BrKNO₃S
  • Sulbactam Related Compound E
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 10.020,00 kr
    10.020,00 kr
    1 * 15 mg
       10.020,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    1623783.
    Beskrivelse
    Sulbactam Related Compound E
    Synonym(s)
    (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide
    CAS-nr
    76646-91-8
    Formel
    C₈H₉Br₂NO₅S
  • Sulbactam Related Compound F
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 9.730,00 kr
    9.730,00 kr
    1 * 15 mg
       9.730,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    1623794.
    Beskrivelse
    Sulbactam Related Compound F
    Synonym(s)
    (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
    CAS-nr
    24158-88-1
    Formel
    C₈H₉Br₂NO₃S