Orders LinkContactUsLinkComponent
Nadolol Related Compounds, Reference Standard
Nadolol Related Compounds, Reference Standard
Leverandør:   USP
Nadolol Related Compounds, Reference Standard
Leverandør:  USP
Certifikater
Nadolol Related Compounds, Reference Standard
5 tilgængelige muligheder nedenfor
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 1449766.
  • 1449711.
  • 1449722.
  • 1449733.
  • 1449777.
  • Beskrivelse:
  • Nadolol Related Compound F
  • Nadolol Related Compound A
  • Nadolol Related Compound B
  • Nadolol Related Compound C
  • Nadolol Related Compound G
  • Synonym(s):
  • 1-(tert-Butylamino)-3-(naphthalen-1-yloxy)propan-2-ol
  • (2RS,3SR)-5-(2,3-Dihydroxypropoxy)-1,2,3,4-tetrahydronaphthalene-2,3-diol
  • (2RS,3SR)-5-(2-Hydroxy-3-methoxypropoxy)-1,2,3,4-tetrahydronaphthalene-2,3-diol
  • 1-(tert-Butylamino)-3-(5,6,7,8-tetrahydronaphthalen-1-yloxy)propan-2-ol
  • CAS-nr:
  • 2116-22-5
  • 33841-03-1
Specifikationer
Product Family Options
Produktinformation
  • Standard
    Pakning
    Pakn
    Tilgængelighed
    Price
  • 1449766.
    Nadolol Related Compound F
    Nadolol Related Compound F
    USP reference standard
    Vial
    25 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 mg
    1 * 25 mg - 9.770,00 kr
    9.770,00 kr
    1 * 25 mg
       9.770,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1449766.
    Beskrivelse
    Nadolol Related Compound F
    Synonym(s)
    1-(tert-Butylamino)-3-(naphthalen-1-yloxy)propan-2-ol
    CAS-nr
    2116-22-5
  • 1449711.
    Nadolol Related Compound A
    Nadolol Related Compound A
    USP reference standard
    Vial
    25 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 mg
    1 * 25 mg - 8.770,00 kr
    8.770,00 kr
    1 * 25 mg
       8.770,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1449711.
    Beskrivelse
    Nadolol Related Compound A
    Synonym(s)
    (2RS,3SR)-5-(2,3-Dihydroxypropoxy)-1,2,3,4-tetrahydronaphthalene-2,3-diol
    CAS-nr
  • 1449722.
    Nadolol Related Compound B
    Nadolol Related Compound B
    USP reference standard
    Vial
    25 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 mg
    1 * 25 mg - 8.770,00 kr
    8.770,00 kr
    1 * 25 mg
       8.770,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1449722.
    Beskrivelse
    Nadolol Related Compound B
    Synonym(s)
    (2RS,3SR)-5-(2-Hydroxy-3-methoxypropoxy)-1,2,3,4-tetrahydronaphthalene-2,3-diol
    CAS-nr
  • 1449733.
    Nadolol Related Compound C
    Nadolol Related Compound C
    USP reference standard
    Vial
    25 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 mg
    1 * 25 mg - 9.210,00 kr
    9.210,00 kr
    1 * 25 mg
       9.210,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1449733.
    Beskrivelse
    Nadolol Related Compound C
    Synonym(s)
    CAS-nr
  • 1449777.
    Nadolol Related Compound G
    Nadolol Related Compound G
    USP reference standard
    Vial
    25 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 mg
    1 * 25 mg - 8.830,00 kr
    8.830,00 kr
    1 * 25 mg
       8.830,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1449777.
    Beskrivelse
    Nadolol Related Compound G
    Synonym(s)
    1-(tert-Butylamino)-3-(5,6,7,8-tetrahydronaphthalen-1-yloxy)propan-2-ol
    CAS-nr
    33841-03-1