Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Losartan Potassium, Reference Standard

Losartan Potassium, Reference Standard

Losartan Potassium, USP reference standard, Vial, 150 mg
Katalog # 1370462.
Leverandør:   USP
Losartan Potassium, Reference Standard
Katalog # 1370462.
Leverandør:  USP
CAS nummer:  
Losartan Potassium, Reference Standard

Losartan Potassium, USP reference standard, Vial, 150 mg

CAS nummer:  
Price
3.140,00 kr
Log ind for tilgængelighed
1 * 150 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Pakn:
    150 mg
  • Standard:
    USP reference standard
  • Beskrivelse:
    Losartan Potassium
  • Cat. No.:
    1370462.
  • Beskrivelse:
    Losartan Potassium
  • CAS-nr:
    124750-99-8
  • Formel:
    C₂₂H₂₂ClKN₆O

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.