Orders LinkContactUsLinkComponent
Betamethasone Acetate, Reference Standards
Betamethasone Acetate, Reference Standards
Leverandør:   USP
Betamethasone Acetate, Reference Standards
Leverandør:  USP
Certifikater
Betamethasone Acetate, Reference Standards
3 tilgængelige muligheder nedenfor
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 1067001.
  • 1067034.
  • 1067045.
  • Beskrivelse:
  • Betamethasone Acetate
  • Betamethasone Acetate Related Compound C
  • Betamethasone Acetate Related Compound D
  • Synonym(s):
  • 9-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 11,21-diacetate
  • 9,11β-Epoxy-17,21-dihydroxy-16β-methylpregna-1,4-diene-3,20-dione 21-acetate
  • CAS-nr:
  • 987-24-6
  • 330157-05-6
  • 912-38-9
  • Formel:
  • C₂₄H₃₁FO₆
  • UN No.:
  • 2811
Specifikationer
Product Family Options
Produktinformation
  • Standard
    Pakning
    Pakn
    Tilgængelighed
    Price
  • 1067001.
    Betamethasone Acetate
    Betamethasone Acetate
    USP reference standard
    Vial
    500 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mg
    1 * 500 mg - 3.140,00 kr
    3.140,00 kr
    1 * 500 mg
       3.140,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1067001.
    Beskrivelse
    Betamethasone Acetate
    Synonym(s)
    CAS-nr
    987-24-6
    Formel
    C₂₄H₃₁FO₆
    UN No.
    2811
  • 1067034.
    Betamethasone Acetate Related Compound C
    Betamethasone Acetate Related Compound C
    USP reference standard
    Vial
    20 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 20 mg
    1 * 20 mg - 18.000,00 kr
    18.000,00 kr
    1 * 20 mg
       18.000,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1067034.
    Beskrivelse
    Betamethasone Acetate Related Compound C
    Synonym(s)
    9-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 11,21-diacetate
    CAS-nr
    330157-05-6
    Formel
    UN No.
  • 1067045.
    Betamethasone Acetate Related Compound D
    Betamethasone Acetate Related Compound D
    USP reference standard
    Vial
    20 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 20 mg
    1 * 20 mg - 25.490,00 kr
    25.490,00 kr
    1 * 20 mg
       25.490,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1067045.
    Beskrivelse
    Betamethasone Acetate Related Compound D
    Synonym(s)
    9,11β-Epoxy-17,21-dihydroxy-16β-methylpregna-1,4-diene-3,20-dione 21-acetate
    CAS-nr
    912-38-9
    Formel
    UN No.
Anbefalinger