Orders LinkContactUsLinkComponent
Topotecan Related Compounds, Reference Standard
 :   USP
Topotecan Related Compounds, Reference Standard
 :  USP
Topotecan Related Compounds, Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard
  • Catalog No:
  • 1672257.
  • 1672268.
  • 1672280.
  • Beskrivelse:
  • Topotecan Hydrochloride
  • Topotecan Related Compound A
  • Topotecan Related Compound C
  • Synonym(s):
  • (S)-10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-8-methoxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
  • (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
  • CAS-nr:
  • 1067286-75-2
  • 7689-03-4
  • UN No.:
  • 1544
  • 1544
Product Family Options
  • Beskrivelse
    Standard
    Pakning
    Pakn
  • Topotecan Hydrochloride
    USP reference standard
    Vial
    350 mg
    1 * 350 mg
    1 * 350 mg - 3.380,00 kr
    3.380,00 kr
    1 * 350 mg
       3.380,00 kr
    Specifications
    Cat. No.
    1672257.
    Beskrivelse
    Topotecan Hydrochloride
    Synonym(s)
    CAS-nr
    1067286-75-2
    UN No.
    1544
  • Topotecan Related Compound A
    USP reference standard
    Vial
    25 mg
    1 * 25 mg
    1 * 25 mg - 4.440,00 kr
    4.440,00 kr
    1 * 25 mg
       4.440,00 kr
    Specifications
    Cat. No.
    1672268.
    Beskrivelse
    Topotecan Related Compound A
    Synonym(s)
    (S)-10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-8-methoxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
    CAS-nr
    UN No.
  • Topotecan Related Compound C
    USP reference standard
    Vial
    25 mg
    1 * 25 mg
    1 * 25 mg - 8.770,00 kr
    8.770,00 kr
    1 * 25 mg
       8.770,00 kr
    Specifications
    Cat. No.
    1672280.
    Beskrivelse
    Topotecan Related Compound C
    Synonym(s)
    (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
    CAS-nr
    7689-03-4
    UN No.
    1544