Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tolyloyl Propionic Acid, Pharmaceutical Analytical Impurities

Tolyloyl Propionic Acid, Pharmaceutical Analytical Impurities

Tolyloyl Propionic Acid, Pharmaceutical Analytical Impurities, Vial, 25 mg
Katalog # 1A01640.
Leverandør:   USP
Tolyloyl Propionic Acid, Pharmaceutical Analytical Impurities
Katalog # 1A01640.
Leverandør:  USP
CAS nummer:  
Tolyloyl Propionic Acid, Pharmaceutical Analytical Impurities

Tolyloyl Propionic Acid, Pharmaceutical Analytical Impurities, Vial, 25 mg

CAS nummer:  
Price
8.140,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Pakn:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • Beskrivelse:
    Tolyloyl Propionic Acid
  • Cat. No.:
    1A01640.
  • Beskrivelse:
    Tolyloyl Propionic Acid
  • CAS-nr:
    4619-20-9
  • Synonym(s):
    4-Oxo-4-(p-tolyl)butanoic Acid

Specifikationer