UA labels 'Clean'
UA labels 'Clean', 95 mm, 95 mm, 500/roll
Catalog # 300-0554
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Ultra-adhesive labels offer a wide range of uses.
- Incorporates a write-on paper front
- High strength, ultra-adhesive for a range of surfaces
- Perfect for dusty surfaces
- Ideal for sealing holes in bags
Ultra-adhesive labels provide a stronger hermetic barrier and can be used for a wide range of applications. The high strength adhesive used means these labels will even stick on very dusty surfaces.
Specifications
- Beskrivelse:UA labels 'Clean'
- Længde:95 mm
- Pakket:500/roll
- Bredde:95 mm