Orders LinkContactUsLinkComponent
VWR®, Total Nitrogen Standards (CRMs)
VWR®, Total Nitrogen Standards (CRMs)
VWR®, Total Nitrogen Standards (CRMs)
Leverandør:  VWR Chemicals
Certifikater
VWR®, Total Nitrogen Standards (CRMs)
VWR®, Total Nitrogen Standards (CRMs)
2 tilgængelige muligheder nedenfor
Product Details & Documents
Certified reference materials for use as standards for Total Nitrogen, typically used as standards for Kjeldahl nitrogen determination.
  • Standard solutions for photometric applications acc EN 25663 (ISO 5663) and ISO 7150
  • Traceable to SRM from NIST
Shelf life: 24 months (12 months after opening)
Certifications: Certified Reference Material acc ISO 17025 / ISO 17034
Product Family Options
Produktinformation
  • Koncentration
    Pakning
    Pakn
    Tilgængelighed
    Price
  • 91273.180
    Total nitrogen standard
    Total nitrogen standard
    100 mg/L
    Plastic bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 632,00 kr
    632,00 kr
    1 * 100 mL
       632,00 kr
  • 91274.180
    Total nitrogen standard
    Total nitrogen standard
    1000 mg/L
    Plastic bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 1.060,00 kr
    1.060,00 kr
    1 * 100 mL
       1.060,00 kr