Orders LinkContactUsLinkComponent
MicroCNX™ connectors
MicroCNX™ connectors
Leverandør:   CPC COLDER
MicroCNX™ connectors
Leverandør:  CPC COLDER
Certifikater
MicroCNX™ connectors
3 tilgængelige muligheder nedenfor
Product Details & Documents
Aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small format biomanufacturing assemblies. MicroCNX™ connectors are the modern alternative to the cumbersome, industrial process of tube welding.
  • Pinch – click and pull: Intuitive three-step connection process reduces risk of operator error
  • Easy to use: Lowers risk of operator error and related performance, reliability and safety concerns
  • Genderless: Eases single-use systems specifications with one part number for both halves
  • CPC click: Audible confirmation of assembly with no additional hardware required
The MicroCNX™ line is engineered specifically for the challenging conditions of biologic media transfer in bioprocessing, cell therapy and gene therapy applications.
Autoclave: One cycle up to 130 °C (266 °F) for 60 minutes
Sterilisation: Up to 50 kGy gamma irradiation
Main components: Polycarbonate (white); USP Class VI, ADCF
Seals: Silicone (clear), platinum-cured; USP Class VI, ADCF
Protective cover: Polypropylene (teal); USP Class VI, ADCF
Membrane: Hydrophobic polyethersulphone; USP Class VI
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 212-0790
  • 212-0792
  • 212-0791
  • Max. operating pressure:
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Max. temperature:
  • 1...40 °C
  • 1...40 °C
  • 1...40 °C
Specifikationer
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • 212-0790
    MicroCNX™ connector, hose barb size 1,6 mm
    MicroCNX™ connector, hose barb size 1,6 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 Items
    1 * 25 Items - 5.930,00 kr
    5.930,00 kr
    1 * 25 Items
       5.930,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    212-0790
    Max. operating pressure
    Up to 4,1 bar; Up to 5,1 bar for 48 hours
    Max. temperature
    1...40 °C
  • 212-0792
    MicroCNX™ connector, hose barb size, 2,4 mm
    MicroCNX™ connector, hose barb size, 2,4 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 Items
    1 * 25 Items - 5.930,00 kr
    5.930,00 kr
    1 * 25 Items
       5.930,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    212-0792
    Max. operating pressure
    Up to 4,1 bar; Up to 5,1 bar for 48 hours
    Max. temperature
    1...40 °C
  • 212-0791
    MicroCNX™ connector, hose barb size, 3,2 mm
    MicroCNX™ connector, hose barb size, 3,2 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 25 Items
    1 * 25 Items - 5.930,00 kr
    5.930,00 kr
    1 * 25 Items
       5.930,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    212-0791
    Max. operating pressure
    Up to 4,1 bar; Up to 5,1 bar for 48 hours
    Max. temperature
    1...40 °C