Orders LinkContactUsLinkComponent
Earmuffs, Peltor™ LiteCom Plus headsets
Earmuffs, Peltor™ LiteCom Plus headsets
Peltor™ LiteCom Plus headset, PMR446 MHz, helmet mounted, 32 dB
Katalog # MMMAMT73H7P3E441EU
Leverandør:   3M
Earmuffs, Peltor™ LiteCom Plus headsets
Katalog # MMMAMT73H7P3E441EU
Leverandør:  3M
Earmuffs, Peltor™ LiteCom Plus headsets
Earmuffs, Peltor™ LiteCom Plus headsets
Peltor™ LiteCom Plus headset, PMR446 MHz, helmet mounted, 32 dB
Price
10.500,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Peltor™ LiteCom plus headsets are robust hearing protectors with a built-in analogue two-way communication radio, noise cancelling dynamic microphone, lithium-ion battery and level dependent functionality for ambient listening.
  • Hearing protector with built-in analogue two-way communication radio
  • Available in 16 channel PMR 446 MHz version with 121 sub-channels
  • Level-dependent technology improves situational awareness and communication
  • Noise cancelling dynamic microphone provides voice-operated transmission for hands-free use
  • External connection jack allows connection of a mobile device or another two-way radio
  • Lithium-ion battery
  • Auto power off and clear battery status indication
  • Available in headband and helmet mounted models
3M™ Peltor™ LiteCom plus headsets feature a built-in analogue two-way radio, available in PMR version with 16 channels and 121 sub-channels, to help users communicate even more efficiently in noisy environments. The noise cancelling dynamic microphone provides voice-operated transmission (VOX) for hands-free use, and an external jack allows connection to a mobile device or another two-way radio.
Sound levels constantly change in demanding situations. When the noise quiets down, turn up the environmental listening function to improve situational awareness or speak with co-workers who are close by, and if the hazardous noise returns the level dependent technology will kick in to help protect user hearing again. Litecom plus headsets have a clear battery status indicator and an auto power off function to save energy.
Specifikationer
  • Hyldehøjde:
    32 dB
  • Beskrivelse:
    Peltor™ LiteCom Plus headset, PMR446 MHz, helmet mounted