Double coated tape, GPT-020F
Katalog # MMMAGPT-020F1540
Leverandør: 3M
Specifikationer
- Bredde:1540 mm
- Farve:transparent
- Beskrivelse:Double coated tape, GPT-020F
- Længde:50 m
- Pakn:1 Roll
Specifikationer
Om denne vare
Designed for versatility, 3M™ double coated tape GPT-020F is a transparent, 200 µm thick, modified solventless acrylic adhesive with a white printed PP filmic liner. It combines a high level of peel and shear performance, enabling high adhesion to a wide variety of substrates.
- Providing the capability to bond to a wide variety of substrates, from high to low surface
- High initial tack and humidity resistance
- Easy handling and converting due to filmic liner delivery
- 200 µm thick
This tape offers high initial tack and humidity resistance, providing easy handling and converting due to the filmic liner delivery. High temperature resistance: 190 °C short term and 90 °C long term.