Orders LinkContactUsLinkComponent
Oxysterol derivatization kit MaxSpec®
Oxysterol derivatization kit MaxSpec®
Oxysterol derivatization MaxSpec® kit
kit contains: Girard's Reagent P, Girard's Reagent P-d5, Oxysterol Derivatization MaxSpec® Cholesterol Oxidase, and Oxysterol Derivatization MaxSpec® System Suitability Mixture, 50 Tests
Katalog # CAYM601540-50
Leverandør:   Cayman Chemical
Oxysterol derivatization kit MaxSpec®
Katalog # CAYM601540-50
Leverandør:  Cayman Chemical
CAS nummer:  
Oxysterol derivatization kit MaxSpec®
Oxysterol derivatization kit MaxSpec®
Oxysterol derivatization MaxSpec® kit
kit contains: Girard's Reagent P, Girard's Reagent P-d5, Oxysterol Derivatization MaxSpec® Cholesterol Oxidase, and Oxysterol Derivatization MaxSpec® System Suitability Mixture, 50 Tests
CAS nummer:  
Price
4.570,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Derivatization Kit for the sensitive LC-MS analysis of oxysterols.
  • Oxidation and derivatization kit for mass spec analysis of oxysterols
  • Includes labelled and unlabelled derivatizing reagents that allow for a variety of sample workflows
  • Can be used for quantitation or untargeted profiling methods
Cayman's Oxysterol Derivatization MaxSpec® Kit is designed for the sensitive detection and relative quantitation of oxysterols in biological samples. This is achieved through enzyme-assisted derivatization utilising d0/d5 labelled Girard’s P reagent to give GP hydrazones. The instrumental response of the derivatized oxysterols is significantly improved due to the presence of a positively charged quaternary nitrogen group in the labelled and unlabelled GP hydrazones. Included in this kit are the essential reagents to derivatize oxysterols in mixtures for sensitive and comparative LC-MS analysis.
Specifikationer
Specifikationer
  • Pakn:
    50 Tests
  • Beskrivelse:
    Oxysterol derivatization MaxSpec® kit<br />kit contains: Girard's Reagent P, Girard's Reagent P-d5, Oxysterol Derivatization MaxSpec® Cholesterol Oxidase, and Oxysterol Derivatization MaxSpec® System Suitability Mixture
Specifikationer