Orders LinkContactUsLinkComponent
Microplate readers, multi-mode, SpectraMax® Mini
Microplate readers, multi-mode, SpectraMax® Mini
Leverandør:   Molecular Devices
Microplate readers, multi-mode, SpectraMax® Mini
Leverandør:  Molecular Devices
Certifikater
Microplate readers, multi-mode, SpectraMax® Mini
Microplate readers, multi-mode, SpectraMax® Mini
12 tilgængelige muligheder nedenfor
Product Details & Documents
The SpectraMax® Mini multi-mode microplate reader offers a budget friendly solution that gives you exactly what you need when you need it. With three modes of detection for UV/Vis absorbance, fluorescence and luminescence, the SpectraMax® Mini reader is user-upgradeable - from two to three modes, providing researchers with a versatile instrument that does not compromise on sensitivity or performance.
  • Industry leading software - enjoy an extensive library of protocols, advanced reader control, and easy data management and analysis with our 'best in class' SoftMax Pro software
  • Approachable design - effortless functionality provides users with everything they need to run their most crucial applications quickly and without complications
  • AutoPMT technology - allows a wide range of concentrations to be prepared and read on one plate, avoiding saturation (or the loss of signal) and saving you time and money
  • Expanded wavelength range - Xenon flash lamps allow you to excite samples in the UV range where halogen does not
  • Affordable - gain access to over 30 years of experience and technology in a compact, multi-mode reader that was built for researchers with sensitive budgets
  • Flexible - perform your favourite applications with a user-friendly reader that accommodates plate types from six to 384-well formats; up to three modes of detection measuring absorbance with NFC-tagged filters for fluorescence, luminescence, or both
  • Easy to use - save time on training, data generation, and data analysis, for faster results
Everything you need to run your most crucial applications quickly and easily.
Access your favourite applications, including ELISA, DNA and protein quantitation, cell viability, gene expression, and more with a compact, upgradeable microplate reader that does everything you need and lets you add detection modes in the future.
The SpectraMax® Mini reader provides tuneable absorbance, filter-based fluorescence and luminescence, temperature control, and easy analysis and data export with SoftMax Pro software.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 735-0395
  • 735-0396
  • 735-0397
  • MLDV5089096
  • MLDV5089097
  • MLDV5089098
  • MLDV5089099
  • MLDV5089100
  • MLDV5089102
  • MLDV5089103
  • MLDV5089104
  • MLDV5089334
  • Temperaturinterval:
  • 4 °C above ambient to 45 °C
  • 4 °C above ambient to 45 °C
  • 4 °C above ambient to 45 °C
  • B×D×H:
  • 400×320×350 mm
  • 400×320×350 mm
  • 400×320×350 mm
  • Vægt:
  • 18 kg
  • 18 kg
  • 18 kg
Specifikationer
Product Family Options
Produktinformation
  • Bølgelængde
    Tilgængelighed
    Price
  • 735-0395
    SpectraMax® Mini multi-mode microplate reader, 2 read modes (absorbance, fluorescence)
    SpectraMax® Mini multi-mode microplate reader, 2 read modes (absorbance, fluorescence)
    200 - 1000 nm (absorbance), 250 - 850 nm (fluorescence)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    735-0395
    Temperaturinterval
    4 °C above ambient to 45 °C
    B×D×H
    400×320×350 mm
    Vægt
    18 kg
  • 735-0396
    SpectraMax® Mini multi-mode microplate reader, 3 read modes (absorbance, fluorescence, luminescence)
    SpectraMax® Mini multi-mode microplate reader, 3 read modes (absorbance, fluorescence, luminescence)
    200 - 1000 nm (absorbance), 250 - 850 nm (fluorescence), 300 - 650 nm (luminescence)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    735-0396
    Temperaturinterval
    4 °C above ambient to 45 °C
    B×D×H
    400×320×350 mm
    Vægt
    18 kg
  • 735-0397
    SpectraMax® Mini multi-mode microplate reader, 2 read modes (absorbance, luminescence)
    SpectraMax® Mini multi-mode microplate reader, 2 read modes (absorbance, luminescence)
    200 - 1000 nm (absorbance), 300 - 650 nm (luminescence)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    735-0397
    Temperaturinterval
    4 °C above ambient to 45 °C
    B×D×H
    400×320×350 mm
    Vægt
    18 kg
Cubes
Produktinformation
  • Bølgelængde
    Tilgængelighed
    Price
  • MLDV5089096
    Assy, standard FL cube, FL-465 (360/405/465)
    Assy, standard FL cube, FL-465 (360/405/465)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089096
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089097
    Assy, standard FL cube, FL-535 (485/508/535)
    Assy, standard FL cube, FL-535 (485/508/535)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089097
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089098
    Assy, standard FL cube, FL-595 (535/555/595)
    Assy, standard FL cube, FL-595 (535/555/595)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089098
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089099
    Assy, optional FL cube, FL-360 (280/310/360)
    Assy, optional FL cube, FL-360 (280/310/360)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089099
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089100
    Assy, optional FL cube, FL-565 (465/508/565)
    Assy, optional FL cube, FL-565 (465/508/565)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089100
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089102
    Assy, optional FL cube, FL-635 (590/610/635)
    Assy, optional FL cube, FL-635 (590/610/635)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089102
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089103
    Assy, optional FL cube, FL-680 (625/655/680)
    Assy, optional FL cube, FL-680 (625/655/680)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089103
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089104
    Assy, optional FL cube, FL-810 (710/760/810)
    Assy, optional FL cube, FL-810 (710/760/810)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089104
    Temperaturinterval
    B×D×H
    Vægt
  • MLDV5089334
    Assy, standard lumi cube
    Assy, standard lumi cube
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    MLDV5089334
    Temperaturinterval
    B×D×H
    Vægt