Multiple affinity removal starter reagent kits
Leverandør: AGILENT
Gå til:
Om denne vare
Starter reagent kit for multiple affinity removal LC columns.
Delivery information: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
4 tilgængelige muligheder nedenfor
Product Details & Documents
Starter reagent kit for multiple affinity removal LC columns.
Delivery information: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
Product Family Options
Produktinformation
- DetailsPaknTilgængelighedPrice
- AGLS5185-5986Multiple affinity removal reagent kit
Components
Produktinformation
- DetailsPaknTilgængelighedPrice
- AGLS5185-5987Multiple affinity removal buffer A, 1 LMultiple affinity removal buffer A, 1 LFor loading, washing, equilibrating1Log ind for tilgængelighed1 * 1 Items993,00 kr
- AGLS5185-5988Multiple affinity removal buffer B, 1 LMultiple affinity removal buffer B, 1 LFor eluting1Log ind for tilgængelighed1 * 1 Items1.342,00 kr
- AGLS5185-5991Spin Concentrators, 4 mlSpin Concentrators, 4 mlto concentrate proteins via low-speed centrifugation through 5K MWCo filters into a collection tube25Log ind for tilgængelighed1 * 25 Items1.968,00 kr