Maybridge Screening library
Om denne vare
The Maybridge portfolio consists of high-quality hit-like, lead-like and drug-like compounds which can generate valuable data in screening programs. They are developed specifically to support the pharmaceutical drug discovery process.
- High-throughput screening for drug discovery
- Maybridge hit-to-lead building blocks
- Maybridge fragment libraries
See our dedicated Maybridge product page for more information and access to full library files, https://uk.vwr.com/cms/maybridge.
: 96-well plates may be provided at mass ≥2 µmol or ≥0,5 mg as powder, dry films, or frozen DMSO solutions. 96- or 384-well plates may be provided at mass 0,1 µmol or 0,01 mg as dry films or frozen DMSO solutions. Compounds may also be ordered in vials at mass 2 µmol or 0,5 mg as powder or DMSO solutions.
We can also provide most major brands of plates and vials and we are happy to use those supplied by customers on request.
8 tilgængelige muligheder nedenfor
Product Details & Documents
The Maybridge portfolio consists of high-quality hit-like, lead-like and drug-like compounds which can generate valuable data in screening programs. They are developed specifically to support the pharmaceutical drug discovery process.
- High-throughput screening for drug discovery
- Maybridge hit-to-lead building blocks
- Maybridge fragment libraries
See our dedicated Maybridge product page for more information and access to full library files, https://uk.vwr.com/cms/maybridge.
: 96-well plates may be provided at mass ≥2 µmol or ≥0,5 mg as powder, dry films, or frozen DMSO solutions. 96- or 384-well plates may be provided at mass 0,1 µmol or 0,01 mg as dry films or frozen DMSO solutions. Compounds may also be ordered in vials at mass 2 µmol or 0,5 mg as powder or DMSO solutions.
We can also provide most major brands of plates and vials and we are happy to use those supplied by customers on request.
Product Family Options
Maybridge HitFinder screening library
Produktinformation
- PaknTilgængelighedPrice
- MAYLHF13384Kit contents: 45 Plates, 0,25 µmol
- MAYLHF13000Kit contents: 180 plates, 1 µmol
Maybridge HitCreater screening library
Produktinformation
- PaknTilgængelighedPrice
- MAYLHC020384Kit contents: 45 Plates, 0,25 µmol
- MAYLHC020000Kit contents: 180 plates, 1 µmol
Maybridge HitDiscover screening library
Produktinformation
- PaknTilgængelighedPrice
- MAYLHD40000Kit contents: 652 plates, 1 µmol
Screening library compounds, hand-selected
Produktinformation
- PaknTilgængelighedPrice
- MAYLHTSPLATEKit contents: Hand-selected 96 well plate
- MAYLFRAGLIB96Kit contents: Hand-selected 96 well, 1 - 5 mg
- MAYLHTSVIALKit contents: Hand-selected, 50×2 ml vial