Orders LinkContactUsLinkComponent
dNTP set
dNTP set
dNTP set, 100 mm each, size: 1 set - 250 µl each, 4
Katalog # 40052.
Leverandør:   Biotium
dNTP set
Katalog # 40052.
Leverandør:  Biotium
CAS nummer:  
dNTP set
dNTP set
dNTP set, 100 mm each, size: 1 set - 250 µl each, 4
CAS nummer:  
Price
1.860,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents
Highly purified dNTPs (>99% by HPLC) in purified water. Each deoxynucleotide solution (dATP, dCTP, dGTP, dTTP) is provided in a separate vial at 100 mm.
The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.
Specifikationer
Specifikationer
  • Pakn:
    4
  • Beskrivelse:
    dNTP set, 100 mm each, size: 1 set - 250 µl each