Masterflex® L/S® Easy-Load® Pump Heads for High-Performance Precision Tubing, Avantor®
Leverandør: Avantor Fluid Handling
Certifikater
Om denne vare
Save time with fast tubing changes and reduced maintenance.
- New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
- Rotor is constructed of stainless steel (SS) for enhanced durability
- Pump heads are stackable for multichannel applications
- Compact design for efficient use of space
- Suitable for use with L/S® High-Performance Precision pump tubing sizes
This Easy-Load® pump head accepts high-performance precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 15 or L/S 24.
- Environmentally Preferable:
Specifikationer
- Catalog No:
- MFLX07514-32
- MFLX07514-12
- Model:
- 07514-32
- 07514-12
- Gear ratio:
- 1:1
- 1:1
- Antal kanaler:
- 1
- 1
- Occlusion:
- Fixed
- Fixed
- Open-head sensor:
- Yes
- No
- Optimum RPM range:
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- Pump series:
- L/S
- L/S
- Suction lift:
- 8,2 m (26,8 ft.)
- 8,2 m (26,8 ft.)
- Tubing retention:
- Automatic
- Automatic
- Tubing sizes accepted:
- L/S® 15, L/S 24
- L/S® 15, L/S 24
- Volume per revolution with L/S 15 tubing:
- 1,7 ml
- 1,7 ml
- Volume per revolution with L/S 24 tubing:
- 2,8 ml
- 2,8 ml
- Garanti:
- 2 years
- 2 years
Specifikationer
Relateret information
Frequently Bought Together