Remote-Control Diaphragm Liquid Dosing Pumps
Om denne vare
Uniformity of discharge ensures low-pulsation transfer process.
- Quiet, reliable, accurate, and virtually maintenance-free
- Pumps can run dry continuously without damage
- Versatile pumps handle flow rates up to 100 ml/min and volumes up to 1000 mL
Simple and intuitive function makes the liquid dosing pump an accurate and reliable solution for your laboratory dosing applications. User-friendly design provides a quiet, dry-running, and self-priming dosing operation. Easy calibration and cycle mode allow for uninterrupted and virtually maintenance-free dosing.
This pump features a PTFE-covered diaphragm and IP65-rated splash protection, making it resistant to chemical attack and adaptable to fluid characteristics.
Unique drive system ensures maximum uniformity of discharge resulting in a low-pulsation transfer process. Pumps feature either local or external control capabilities including start/stop and reset/prime via TTL contact as well as output signal fault alarm. Models available with a RS-232 port for connectivity.
: Includes 5-ft (1.5-m) power cord and two 3-ft (1-m) pieces of FEP inlet/outlet tubing (1/8" OD x 1/16" ID) with UNF 1/4"-28 fittings. Models with RS-232 port also include a cable with D-Sub-9 connector.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Uniformity of discharge ensures low-pulsation transfer process.
- Quiet, reliable, accurate, and virtually maintenance-free
- Pumps can run dry continuously without damage
- Versatile pumps handle flow rates up to 100 ml/min and volumes up to 1000 mL
Simple and intuitive function makes the liquid dosing pump an accurate and reliable solution for your laboratory dosing applications. User-friendly design provides a quiet, dry-running, and self-priming dosing operation. Easy calibration and cycle mode allow for uninterrupted and virtually maintenance-free dosing.
This pump features a PTFE-covered diaphragm and IP65-rated splash protection, making it resistant to chemical attack and adaptable to fluid characteristics.
Unique drive system ensures maximum uniformity of discharge resulting in a low-pulsation transfer process. Pumps feature either local or external control capabilities including start/stop and reset/prime via TTL contact as well as output signal fault alarm. Models available with a RS-232 port for connectivity.
: Includes 5-ft (1.5-m) power cord and two 3-ft (1-m) pieces of FEP inlet/outlet tubing (1/8" OD x 1/16" ID) with UNF 1/4"-28 fittings. Models with RS-232 port also include a cable with D-Sub-9 connector.
Specifications for Product Family
Specifikationer
- Catalog No:
- MFLX78167-09
- MFLX78167-01
- Inlet connection:
- 5/16" hose barb
- 5/16" hose barb
- Maks. tryk:
- 5,86 bar (85 psi)
- 5,86 bar (85 psi)
- Max. temperature:
- 80 °C (176 °F)
- 80 °C (176 °F)
- Tilslutning af stikkontakt:
- 5/16" hose barb
- 5/16" hose barb
- Power cord:
- 6-ft power cord with 3-prong U.S. plug
- 6-ft power cord with 3-prong U.S. plug
- Run dry:
- Yes
- Yes
- Suction lift, wet:
- 3962 mm of H₂O (156" H₂O)
- 3962 mm of H₂O (156" H₂O)
- Våde materialer:
- PVDF head, PTFE diaphragm, FFKM valves
- PVDF head, PTFE diaphragm, FFKM valves
- Garanti:
- 1 year
- 1 year
Specifikationer
Product Family Options
Produktinformation
- Maksimal flowhastighedPowerHøjdeLængdeBreddeTilgængelighedPrice
- MFLX78167-09Digital Remote Control Dosing Pump, PVDF head, 1 to 100 ml/minDigital Remote Control Dosing Pump, PVDF head, 1 to 100 ml/min100 ml/min100 - 230 VAC; 50/60 Hz; 0,54 A19,1 cm15,9 cm10,8 cmLog ind for tilgængelighed1 * 1 Items21.490,00 krSpecifikationer:CE CompliantIP65 washdown
More
Specifications
Cat. No.MFLX78167-09Inlet connection5/16" hose barbMaks. tryk5,86 bar (85 psi)Max. temperature80 °C (176 °F)Tilslutning af stikkontakt5/16" hose barbPower cord6-ft power cord with 3-prong U.S. plugRun dryYesSuction lift, wet3962 mm of H₂O (156" H₂O)Våde materialerPVDF head, PTFE diaphragm, FFKM valvesGaranti1 yearCE CompliantIP65 washdown - MFLX78167-01Digital Remote Control Dosing Pump, PVDF head, 0,03 to 20 ml/minDigital Remote Control Dosing Pump, PVDF head, 0,03 to 20 ml/min20 ml/min100 - 230 VAC; 50/60 Hz; 0,54 A19,1 cm15,9 cm10,8 cmLog ind for tilgængelighed1 * 1 Items23.280,00 krSpecifikationer:CE CompliantIP65 washdown
More
Specifications
Cat. No.MFLX78167-01Inlet connection5/16" hose barbMaks. tryk5,86 bar (85 psi)Max. temperature80 °C (176 °F)Tilslutning af stikkontakt5/16" hose barbPower cord6-ft power cord with 3-prong U.S. plugRun dryYesSuction lift, wet3962 mm of H₂O (156" H₂O)Våde materialerPVDF head, PTFE diaphragm, FFKM valvesGaranti1 yearCE CompliantIP65 washdown
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- In-line Filter 70um PVDF for Dosing Pumps 78167-08 to -10In-line Filter 70um PVDF for Dosing Pumps 78167-08 to -10MFLX78167-52Log ind for tilgængelighed1 * 1 Items - 1.360,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 FittingConnection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 FittingMFLX78167-55Log ind for tilgængelighed1 * 1 Items - 1.030,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 FittingConnection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 FittingMFLX78167-56Log ind for tilgængelighed1 * 1 Items - 1.830,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Chemicals Valve Kit for Dosing Pumps 78167-00 to -10Chemicals Valve Kit for Dosing Pumps 78167-00 to -10MFLX78167-63Log ind for tilgængelighed1 * 1 Items - 5.010,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07MFLX78167-50Log ind for tilgængelighed1 * 1 Items - 1.040,00 kr