Masterflex® Pressure Booster Pump Systems, Avantor®
Om denne vare
Pump system automatically adjusts speed to maintain set pressure.
- Maximum flow range of 25 to 100 GPM│NEMA 1 or NEMA 4 variable frequency drive controller with corresponding main disconnect
- Three-phase input line voltage (single-phase systems are available)
Pump supply booster systems feature a stainless steel centrifugal pump. System is controlled by the variable frequency drive (VFD) which features an LCD and adjustable pressure range; choose the VFD with either a NEMA 1 or NEMA 4 rated enclosure and main disconnect.
System has a small footprint of approximately 30" x 30" (76.2 x 76.2 cm), and has a 1.5" NPT(F) suction and discharge connection. Piping, valve, and drain bodies are made from brass to meet most needs.
Additional options are available: stainless steel piping upgrade, suction check valve (bronze), unions (bronze), low suction shutoff, and low suction shutoff with alarm. If your facility has only single phase voltage, a custom unit can be made to match. Please contact our Application Specialists for details.
Please contact your local Avantor office to customize a system suited to your needs.
: Includes pump, VFD controller, bladder tank, suction and discharge gauges, discharge check valve, backup relief valve, and necessary piping and drain.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Pump system automatically adjusts speed to maintain set pressure.
- Maximum flow range of 25 to 100 GPM│NEMA 1 or NEMA 4 variable frequency drive controller with corresponding main disconnect
- Three-phase input line voltage (single-phase systems are available)
Pump supply booster systems feature a stainless steel centrifugal pump. System is controlled by the variable frequency drive (VFD) which features an LCD and adjustable pressure range; choose the VFD with either a NEMA 1 or NEMA 4 rated enclosure and main disconnect.
System has a small footprint of approximately 30" x 30" (76.2 x 76.2 cm), and has a 1.5" NPT(F) suction and discharge connection. Piping, valve, and drain bodies are made from brass to meet most needs.
Additional options are available: stainless steel piping upgrade, suction check valve (bronze), unions (bronze), low suction shutoff, and low suction shutoff with alarm. If your facility has only single phase voltage, a custom unit can be made to match. Please contact our Application Specialists for details.
Please contact your local Avantor office to customize a system suited to your needs.
: Includes pump, VFD controller, bladder tank, suction and discharge gauges, discharge check valve, backup relief valve, and necessary piping and drain.
Specifications for Product Family
Specifikationer
- Catalog No:
- MFLX70794-44
- MFLX70794-64
- Model:
- 70794-44
- 70794-64
- Maks. tryk:
- 4,3 bar (63 psi)
- 3,9 bar (56 psi)
- Motor enclosure type:
- NEMA 4
- NEMA 4
- Motor hp:
- 2
- 3
- Våde materialer:
- SS pump, brass piping/valves/drain
- SS pump, brass piping/valves/drain
- Garanti:
- 1 year
- 1 year
Specifikationer
Product Family Options
Produktinformation
- Maksimal flowhastighedTilgængelighedPrice
- MFLX70794-44Pump Booster System, 38 gal./min or 63 PSI, 2 HP, NEMA 4 enclosurePump Booster System, 38 gal./min or 63 PSI, 2 HP, NEMA 4 enclosure143846 ml/min (38 gal./min)Log ind for tilgængelighed1 * 1 Items43.200,00 krSpecifikationer:CE Compliant
More
Specifications
Cat. No.MFLX70794-44Model70794-44Maks. tryk4,3 bar (63 psi)Motor enclosure typeNEMA 4Motor hp2Våde materialerSS pump, brass piping/valves/drainGaranti1 yearCE Compliant - MFLX70794-64Pump Booster System, 100 gal./min or 56 PSI, 3 HP, NEMA 4 enclosurePump Booster System, 100 gal./min or 56 PSI, 3 HP, NEMA 4 enclosure378541 ml/min (100 gal./min)Log ind for tilgængelighed1 * 1 Items45.290,00 krSpecifikationer:CE Compliant
More
Specifications
Cat. No.MFLX70794-64Model70794-64Maks. tryk3,9 bar (56 psi)Motor enclosure typeNEMA 4Motor hp3Våde materialerSS pump, brass piping/valves/drainGaranti1 yearCE Compliant