Masterflex® Sanitary Gaskets, Polymers, Avantor®
Om denne vare
Ensure a tight seal.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimise downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
32 tilgængelige muligheder nedenfor
Product Details & Documents
Ensure a tight seal.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimise downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
Specifications for Product Family
Specifikationer
- Catalog No:
- MFLX30548-40
- MFLX30548-42
- MFLX30548-44
- MFLX30548-46
- MFLX30548-48
- MFLX29590-80
- MFLX30548-60
- MFLX30548-62
- MFLX30548-64
- MFLX30548-66
- MFLX30548-68
- MFLX29590-83
- MFLX00162-NJ
- MFLX00364-YQ
- MFLX00404-MO
- MFLX30548-00
- MFLX30548-02
- MFLX30548-04
- MFLX30548-06
- MFLX30548-08
- MFLX41001-77
- MFLX30548-80
- MFLX30548-82
- MFLX30548-84
- MFLX30548-86
- MFLX30548-88
- MFLX16011-66
- MFLX30548-20
- MFLX30548-24
- MFLX30548-26
- MFLX30548-28
- MFLX30548-22
- Connection type:
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Garanti:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifikationer
Product Family Options
Produktinformation
- Fitting materialSanitary clamp size (Port 1)Sanitary gasket sizeTilgængelighedPrice
- MFLX30548-40Sanitary Gasket, Buna-N, 3/4" Tri-Clamp®; 10/PKSanitary Gasket, Buna-N, 3/4" Tri-Clamp®; 10/PKBuna-N3/4"3/4"Log ind for tilgængelighed1 * 10 Items186,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX30548-40Connection typeSanitaryGaranti1 year - MFLX30548-42Sanitary Gasket, Buna-N, 1" Tri-Clamp®; 10/PKSanitary Gasket, Buna-N, 1" Tri-Clamp®; 10/PKBuna-N1-1/2"1"Log ind for tilgængelighed1 * 10 Items91,80 krSpecifikationer:
More
Specifications
Cat. No.MFLX30548-42Connection typeSanitaryGaranti1 year - MFLX30548-44Sanitary Gasket, Buna-N, 1-1/2" Tri-Clamp®; 10/PKSanitary Gasket, Buna-N, 1-1/2" Tri-Clamp®; 10/PKBuna-N1-1/2"1-1/2"Log ind for tilgængelighed1 * 10 Items88,10 krSpecifikationer:
More
Specifications
Cat. No.MFLX30548-44Connection typeSanitaryGaranti1 year - MFLX30548-46Sanitary Gasket, Buna-N, 2" Tri-Clamp®; 10/PKSanitary Gasket, Buna-N, 2" Tri-Clamp®; 10/PKBuna-N2"2"Log ind for tilgængelighed1 * 10 Items81,40 krSpecifikationer:
More
Specifications
Cat. No.MFLX30548-46Connection typeSanitaryGaranti1 year - MFLX30548-48Sanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PKSanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PKBuna-N2"2-1/2"Log ind for tilgængelighed1 * 10 Items97,80 krSpecifikationer:
More
Specifications
Cat. No.MFLX30548-48Connection typeSanitaryGaranti1 year - MFLX29590-80Buna N Sanitary Gasket, Black, 4" Tri-Clamp; 1/PkBuna N Sanitary Gasket, Black, 4" Tri-Clamp; 1/PkBuna N4"4"Log ind for tilgængelighed1 * 1 Items34,40 krSpecifikationer:
More
Specifications
Cat. No.MFLX29590-80Connection typeSanitaryGaranti1 year - MFLX30548-60Sanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PKSanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PKEPDM3/4"3/4"Log ind for tilgængelighed1 * 10 Items191,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-60Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-62Sanitary Gasket, EPDM, 1" Tri-Clamp®; 10/PKSanitary Gasket, EPDM, 1" Tri-Clamp®; 10/PKEPDM1-1/2"1"Log ind for tilgængelighed1 * 10 Items197,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX30548-62Connection typeSanitaryGaranti1 year - MFLX30548-64Sanitary Gasket, EPDM, 1-1/2" Tri-Clamp®; 10/PKSanitary Gasket, EPDM, 1-1/2" Tri-Clamp®; 10/PKEPDM1-1/2"1-1/2"Log ind for tilgængelighed1 * 10 Items187,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-64Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-66Sanitary Gasket, EPDM, 2" Tri-Clamp®; 10/PKSanitary Gasket, EPDM, 2" Tri-Clamp®; 10/PKEPDM2"2"Log ind for tilgængelighed1 * 10 Items201,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-66Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-68Sanitary Gasket, EPDM, 2-1/2" Tri-Clamp®; 10/PKSanitary Gasket, EPDM, 2-1/2" Tri-Clamp®; 10/PKEPDM2"2-1/2"Log ind for tilgængelighed1 * 10 Items214,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-68Connection typeSanitaryGaranti1 yearCE certificate - MFLX29590-83FKM/Viton Sanitary Gasket, 4" Tri-Clamp; 1/PkFKM/Viton Sanitary Gasket, 4" Tri-Clamp; 1/PkFKM/Viton4"4"Log ind for tilgængelighed1 * 1 Items144,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX29590-83Connection typeSanitaryGaranti1 year - MFLX00162-NJPlatinum-Cured Silicone Gasket, White, 2"; 10/PKPlatinum-Cured Silicone Gasket, White, 2"; 10/PKPlatinum-Cured Silicone2"2"Log ind for tilgængelighed1 * 10 Items184,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX00162-NJConnection typeSanitaryGaranti1 year - MFLX00364-YQSanitary Gasket, Platinum-Cured Silicone, 4"; 10/PKSanitary Gasket, Platinum-Cured Silicone, 4"; 10/PKPlatinum-Cured Silicone4"4"Log ind for tilgængelighed1 * 10 Items514,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX00364-YQConnection typeSanitaryGaranti1 year - MFLX00404-MOSanitary Gasket, Platinum-Cured Silicone, 6"; 10/PKSanitary Gasket, Platinum-Cured Silicone, 6"; 10/PKPlatinum-Cured Silicone6"6"Log ind for tilgængelighed1 * 10 Items3.060,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX00404-MOConnection typeSanitaryGaranti1 year - MFLX30548-00Sanitary Gasket, Silicone, 3/4" Tri-Clamp®; 10/PKSanitary Gasket, Silicone, 3/4" Tri-Clamp®; 10/PKPlatinum-Cured Silicone3/4"3/4"Log ind for tilgængelighed1 * 10 Items197,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-00Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-02Sanitary Gasket, Silicone, 1"; 10/PKSanitary Gasket, Silicone, 1"; 10/PKPlatinum-Cured Silicone1-1/2"1"Log ind for tilgængelighed1 * 10 Items271,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-02Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-04Sanitary Gasket, Silicone, 1-1/2"; 10/PKSanitary Gasket, Silicone, 1-1/2"; 10/PKPlatinum-Cured Silicone1-1/2"1-1/2"Log ind for tilgængelighed1 * 10 Items197,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-04Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-06Sanitary Gasket, Silicone, 2"; 10/PKSanitary Gasket, Silicone, 2"; 10/PKPlatinum-Cured Silicone2"2"Log ind for tilgængelighed1 * 10 Items271,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-06Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-08Sanitary Gasket, Silicone, 2-1/2"; 10/PKSanitary Gasket, Silicone, 2-1/2"; 10/PKPlatinum-Cured Silicone2"2-1/2"Log ind for tilgængelighed1 * 10 Items299,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-08Connection typeSanitaryGaranti1 yearCE certificate - MFLX41001-77Sanitary Gasket, Platinum-Cured Silicone, 3"; 10/PKSanitary Gasket, Platinum-Cured Silicone, 3"; 10/PKPlatinum-Cured Silicone3"3"Log ind for tilgængelighed1 * 10 Items344,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX41001-77Connection typeSanitaryGaranti1 year - MFLX30548-80Sanitary Gasket, PTFE, 3/4" Tri-Clamp®; 10/PKSanitary Gasket, PTFE, 3/4" Tri-Clamp®; 10/PKPTFE3/4"3/4"Log ind for tilgængelighed1 * 10 Items235,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-80Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-82Sanitary Gasket, PTFE, 1" Tri-Clamp®; 10/PKSanitary Gasket, PTFE, 1" Tri-Clamp®; 10/PKPTFE1-1/2"1"Log ind for tilgængelighed1 * 10 Items309,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-82Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-84Sanitary Gasket, PTFE, 1-1/2" Tri-Clamp®; 10/PKSanitary Gasket, PTFE, 1-1/2" Tri-Clamp®; 10/PKPTFE1-1/2"1-1/2"Log ind for tilgængelighed1 * 10 Items286,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-84Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-86Sanitary Gasket, PTFE, 2" Tri-Clamp®; 10/PKSanitary Gasket, PTFE, 2" Tri-Clamp®; 10/PKPTFE2"2"Log ind for tilgængelighed1 * 10 Items299,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-86Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-88Sanitary Gasket, PTFE, 2-1/2" Tri-Clamp®; 10/PKSanitary Gasket, PTFE, 2-1/2" Tri-Clamp®; 10/PKPTFE2"2-1/2"Log ind for tilgængelighed1 * 10 Items328,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-88Connection typeSanitaryGaranti1 yearCE certificate - MFLX16011-66Silicone Sanitary Gasket, 0,5" Tri-Clamp; 10/PkSilicone Sanitary Gasket, 0,5" Tri-Clamp; 10/PkSilicone3/4"0,5"Log ind for tilgængelighed1 * 10 Items206,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX16011-66Connection typeSanitaryGaranti1 year - MFLX30548-20Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PKSanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PKViton®3/4"3/4"Log ind for tilgængelighed1 * 10 Items386,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-20Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-24Sanitary Gasket, Viton®, 1-1/2" Tri-Clamp®; 10/PKSanitary Gasket, Viton®, 1-1/2" Tri-Clamp®; 10/PKViton®1-1/2"1-1/2"Log ind for tilgængelighed1 * 10 Items381,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-24Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-26Sanitary Gasket, Viton®, 2" Tri-Clamp®; 10/PKSanitary Gasket, Viton®, 2" Tri-Clamp®; 10/PKViton®2"2"Log ind for tilgængelighed1 * 10 Items393,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-26Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-28Sanitary Gasket, Viton®, 2-1/2" Tri-Clamp®; 10/PKSanitary Gasket, Viton®, 2-1/2" Tri-Clamp®; 10/PKViton®2"2-1/2"Log ind for tilgængelighed1 * 10 Items456,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-28Connection typeSanitaryGaranti1 yearCE certificate - MFLX30548-22Sanitary Gasket, Viton®, 1" Tri-Clamp®; 10/PKSanitary Gasket, Viton®, 1" Tri-Clamp®; 10/PKViton®1-1/2"1"Log ind for tilgængelighed1 * 10 Items424,00 krSpecifikationer:CE certificate
More
Specifications
Cat. No.MFLX30548-22Connection typeSanitaryGaranti1 yearCE certificate