Explosion-Proof NEMA Type C-Face Motors 47718
Leverandør: Avantor Fluid Handling
Certifikater
Om denne vare
For Class 1 Group C and D, Class 2 Group F and G locations.
- Automatic overload protection stops and restarts motor in case of overload or overheating
- UL-listed file no. E-12044
Motors are TEFC (models 70007-00 to -12 are TENV) and include an explosion-proof conduit box (shipped loose).
- Environmentally Preferable:
Specifikationer
- Catalog No:
- MFLX71006-45
- MFLX71006-05
- Motor enclosure type:
- TEFC
- TEFC
- Power conduit box connection:
- Yes
- Yes
- Garanti:
- 1 year
- 1 year