Masterflex® Transfer Tubing Spool, Polytetrafluoroethylene (PTFE), Avantor®
Leverandør: Avantor Fluid Handling
Certifikater
Om denne vare
Available in spooled rolls for increased versatility.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.
- Environmentally Preferable:
Specifikationer
- Model:
- 95231-06
- 95231-02
- 95231-00
- 95231-08
- 95231-10
- 95231-12
- 95231-14
- Bøjningsradius:
- 25,4 mm (1")
- 12,7 mm (0,5")
- 12,7 mm (0,5")
- 44,5 mm (1,75")
- 25,4 mm (1")
- 63,5 mm (2,5")
- 50,8 mm (2")
- Clarity:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 30,5 m (100 ft.)
- Maks. tryk:
- 9,9 bar (144 psi)
- 23,4 bar (339 psi)
- 23,2 bar (336 psi)
- 7,9 bar (114 psi)
- 14,1 bar (204 psi)
- 6,3 bar (92 psi)
- 5,4 bar (79 psi)
- Max. temperature:
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- Min. temperature:
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- Regulatory approvals:
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- Working pressure:
- 9,9 bar (144 psi)
- 23,4 bar (339 psi)
- 23,2 bar (336 psi)
- 7,9 bar (114 psi)
- 14,1 bar (204 psi)
- 6,3 bar (92 psi)
- 5,4 bar (79 psi)
- Garanti:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year