Masterflex® Transfer Tubing Spool, Fluorinated Ethylene Propylene (FEP), Avantor®
Leverandør: Avantor Fluid Handling
Certifikater
Om denne vare
Maximize tubing on hand.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.
- Environmentally Preferable:
Specifikationer
- Catalog No:
- MFLX95230-00
- MFLX95230-04
- MFLX95230-06
- MFLX95230-12
- MFLX95230-14
- MFLX95230-08
- MFLX95230-10
- MFLX95230-02
- Model:
- 95230-00
- 95230-04
- 95230-06
- 95230-12
- 95230-14
- 95230-08
- 95230-10
- 95230-02
- Bøjningsradius:
- 12,7 mm (0,5")
- 19,1 mm (0,75")
- 25,4 mm (1")
- 63,5 mm (2,5")
- 50,8 mm (2")
- 44,5 mm (1,75")
- 25,5 mm (1")
- 12,7 mm (0,5")
- Clarity:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 30,5 m (100 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- 76,2 m (250 ft.)
- Maks. tryk:
- 28,4 bar (412 psi)
- 19,9 bar (289 psi)
- 12,2 bar (177 psi)
- 7,8 bar (113 psi)
- 6,7 bar (97 psi)
- 9,7 bar (140 psi)
- 17,2 bar (250 psi)
- 28,75 bar (417 psi)
- Max. temperature:
- 260 °C (500 °F)
- 260 °C (500 °F)
- 260 °C (500 °F)
- 260 °C (500 °F)
- 260 °C (500 °F)
- 260 °C (500 °F)
- 260 °C (500 °F)
- 260 °C (500 °F)
- Min. temperature:
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- Regulatory approvals:
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- Working pressure:
- 28,4 bar (412 psi)
- 19,9 bar (289 psi)
- 12,2 bar (177 psi)
- 7,8 bar (113 psi)
- 6,7 bar (97 psi)
- 9,7 bar (140 psi)
- 17,2 bar (250 psi)
- 28,75 bar (417 psi)
- Garanti:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year