About this item
A 'must have' personal tool for every laboratory-ideal for quick spin.
- Small foot print with rubber feet keep unit stable
- Incudes two rotors: one for 1,5/2,0 ml tubes and one for PCR tubes
- On/off switch lets you stop rotor before opening lid
These mini microcentrifuges offer a choice of five translucent lid colors. Operating is extremely simple: just close the lid and the rotor accelerates quicly to 6000 rpm. Microcentrifuges include two sets of adapters for 0,4 to 0,5 ml tubes.
- Environmentally Preferable:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.