Masterflex® Transfer Tubing, Ester-Based Polyurethane, Avantor®
Om denne vare
Worry-free petroleum-based transfer.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
The tubes are ideal for high-purity, general laboratory, petroleum-based transfer. Characteristics include plasticizer-free, low extractables, good at low temperatures, good resistance to petroleum-based chemicals, and high tensile strength. Not recommended for use with aqueous solutions. Temperature range is -70 to 185 °F (-56 to 85 °C). Can be sterilized with ethylene oxide.
17 tilgængelige muligheder nedenfor
Product Details & Documents
Worry-free petroleum-based transfer.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
The tubes are ideal for high-purity, general laboratory, petroleum-based transfer. Characteristics include plasticizer-free, low extractables, good at low temperatures, good resistance to petroleum-based chemicals, and high tensile strength. Not recommended for use with aqueous solutions. Temperature range is -70 to 185 °F (-56 to 85 °C). Can be sterilized with ethylene oxide.
Specifications for Product Family
Specifikationer
- Catalog No:
- MFLX95680-36
- MFLX95680-32
- MFLX95680-14
- MFLX95680-12
- MFLX95680-16
- MFLX95680-04
- MFLX95680-02
- MFLX95680-00
- MFLX95680-50
- MFLX95680-46
- MFLX95680-22
- MFLX95680-26
- MFLX95680-06
- MFLX95680-08
- MFLX95680-42
- MFLX95680-20
- MFLX95680-28
- Model:
- 95680-36
- 95680-32
- 95680-14
- 95680-12
- 95680-16
- 95680-04
- 95680-02
- 95680-00
- 95680-50
- 95680-46
- 95680-22
- 95680-26
- 95680-06
- 95680-08
- 95680-42
- 95680-20
- 95680-28
- Bøjningsradius:
- 44,45 mm (1,75")
- 73,025 mm (2,875")
- 23,8125 mm (0,9375")
- 39,6875 mm (1,5625")
- 17,4625 mm (0,6875")
- 7,9375 mm (0,3125")
- 12,7 mm (0,5")
- 4,7625 mm (0,1875")
- 130,175 mm (5,125")
- 82,55 mm (3,25")
- 44,45 mm (1,75")
- 26,9875 mm (1,0625")
- 25,4 mm (1")
- 15,875 mm (0,625")
- 60,325 mm (2,375")
- 49,2125 mm (1,9375")
- 42,8625 mm (1,6875")
- Clarity:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- Maks. tryk:
- 4,1 bar (60 psi)
- 2,5 bar (36 psi)
- 4,6 bar (67 psi)
- 2,3 bar (33 psi)
- 5,1 bar (74 psi)
- 6,9 bar (100 psi)
- 5 bar (72 psi)
- 6,9 bar (100 psi)
- 2,8 bar (40 psi)
- 3,5 bar (51 psi)
- 4,1 bar (59 psi)
- 6,9 bar (100 psi)
- 1,9 bar (27 psi)
- 5,1 bar (74 psi)
- 4,1 bar (60 psi)
- 4,5 bar (65 psi)
- 5,1 bar (74 psi)
- Max. temperature:
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- Max. vacuum:
- 29,9" Hg
- 20" Hg
- 29,9" Hg
- 20" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 15" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- Min. temperatur:
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Working pressure:
- 4,1 bar (60 psi)
- 2,5 bar (36 psi)
- 4,6 bar (67 psi)
- 2,3 bar (33 psi)
- 5,1 bar (74 psi)
- 6,9 bar (100 psi)
- 5 bar (72 psi)
- 6,9 bar (100 psi)
- 2,8 bar (40 psi)
- 3,5 bar (51 psi)
- 4,1 bar (59 psi)
- 6,9 bar (100 psi)
- 1,9 bar (27 psi)
- 5,1 bar (74 psi)
- 4,1 bar (60 psi)
- 4,5 bar (65 psi)
- 5,1 bar (74 psi)
Specifikationer
Product Family Options
Produktinformation
- Rør int.ØVægtykkelseRør udv.ØSlangematerialeLængdeFarveTilgængelighedPrice
- MFLX95680-36Transfer Tubing, Ester-Based Polyurethane, 1/2" ID x 3/4" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/2" ID x 3/4" OD; 100 Ft12,8 mm (0,5")3,2 mm19,2 mm (0,75")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod6.130,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-36Model95680-36Bøjningsradius44,45 mm (1,75")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk4,1 bar (60 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4,1 bar (60 psi) - MFLX95680-32Transfer Tubing, Ester-Based Polyurethane, 1/2" ID x 5/8" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/2" ID x 5/8" OD; 100 Ft12,8 mm (0,5")1,6 mm16 mm (0,625")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod3.150,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-32Model95680-32Bøjningsradius73,025 mm (2,875")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk2,5 bar (36 psi)Max. temperature93 °C (200 °F)Max. vacuum20" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure2,5 bar (36 psi) - MFLX95680-14Transfer Tubing, Ester-Based Polyurethane, 1/4" ID x 3/8" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/4" ID x 3/8" OD; 100 Ft6,4 mm (0,25")1,6 mm9,6 mm (0,375")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod1.960,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-14Model95680-14Bøjningsradius23,8125 mm (0,9375")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk4,6 bar (67 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4,6 bar (67 psi) - MFLX95680-12Transfer Tubing, Ester-Based Polyurethane, 1/4" ID x 5/16" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/4" ID x 5/16" OD; 100 Ft6,4 mm (0,25")0,8 mm8 mm (0,3125")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod1.680,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-12Model95680-12Bøjningsradius39,6875 mm (1,5625")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk2,3 bar (33 psi)Max. temperature93 °C (200 °F)Max. vacuum20" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure2,3 bar (33 psi) - MFLX95680-16Transfer Tubing, Ester-Based Polyurethane, 1/4" ID x 7/16" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/4" ID x 7/16" OD; 100 Ft6,4 mm (0,25")2,4 mm11,2 mm (0,4375")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod3.510,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-16Model95680-16Bøjningsradius17,4625 mm (0,6875")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk5,1 bar (74 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5,1 bar (74 psi) - MFLX95680-04Transfer Tubing, Ester-Based Polyurethane, 1/8" ID x 1/4" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/8" ID x 1/4" OD; 100 Ft3,2 mm (0,125")1,6 mm6,4 mm (0,25")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod1.370,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-04Model95680-04Bøjningsradius7,9375 mm (0,3125")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk6,9 bar (100 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure6,9 bar (100 psi) - MFLX95680-02Transfer Tubing, Ester-Based Polyurethane, 1/8" ID x 3/16" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/8" ID x 3/16" OD; 100 Ft3,2 mm (0,125")0,8 mm4,8 mm (0,1875")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod720,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-02Model95680-02Bøjningsradius12,7 mm (0,5")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk5 bar (72 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5 bar (72 psi) - MFLX95680-00Transfer Tubing, Ester-Based Polyurethane, 1/16" ID x 1/8" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1/16" ID x 1/8" OD; 100 Ft1,6 mm (0,0625")0,8 mm3,2 mm (0,125")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod587,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-00Model95680-00Bøjningsradius4,7625 mm (0,1875")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk6,9 bar (100 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure6,9 bar (100 psi) - MFLX95680-50Transfer Tubing, Ester-Based Polyurethane, 1" ID x 1-1/4" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 1" ID x 1-1/4" OD; 100 Ft25,4 mm (1")3,2 mm31,8 mm (1,25")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod10.900,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-50Model95680-50Bøjningsradius130,175 mm (5,125")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk2,8 bar (40 psi)Max. temperature93 °C (200 °F)Max. vacuum15" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure2,8 bar (40 psi) - MFLX95680-46Transfer Tubing, Ester-Based Polyurethane, 3/4" ID x 1" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 3/4" ID x 1" OD; 100 Ft19,2 mm (0,75")3,2 mm25,4 mm (1")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod7.290,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-46Model95680-46Bøjningsradius82,55 mm (3,25")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk3,5 bar (51 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure3,5 bar (51 psi) - MFLX95680-22Transfer Tubing, Ester-Based Polyurethane, 3/8" ID x 1/2" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 3/8" ID x 1/2" OD; 100 Ft9,6 mm (0,375")1,6 mm12,8 mm (0,5")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod2.460,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-22Model95680-22Bøjningsradius44,45 mm (1,75")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk4,1 bar (59 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4,1 bar (59 psi) - MFLX95680-26Transfer Tubing, Ester-Based Polyurethane, 3/8" ID x 5/8" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 3/8" ID x 5/8" OD; 100 Ft9,6 mm (0,375")3,2 mm16 mm (0,625")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod4.680,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-26Model95680-26Bøjningsradius26,9875 mm (1,0625")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk6,9 bar (100 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure6,9 bar (100 psi) - MFLX95680-06Transfer Tubing, Ester-Based Polyurethane, 3/16" ID x 1/4" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 3/16" ID x 1/4" OD; 100 Ft4,8 mm (0,1875")0,8 mm6,4 mm (0,25")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod918,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-06Model95680-06Bøjningsradius25,4 mm (1")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk1,9 bar (27 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure1,9 bar (27 psi) - MFLX95680-08Transfer Tubing, Ester-Based Polyurethane, 3/16" ID x 5/16" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 3/16" ID x 5/16" OD; 100 Ft4,8 mm (0,1875")1,6 mm8 mm (0,3125")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod1.650,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-08Model95680-08Bøjningsradius15,875 mm (0,625")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk5,1 bar (74 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5,1 bar (74 psi) - MFLX95680-42Transfer Tubing, Ester-Based Polyurethane, 5/8" ID x 7/8" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 5/8" ID x 7/8" OD; 100 Ft16 mm (0,625")3,2 mm22,4 mm (0,875")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod6.660,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-42Model95680-42Bøjningsradius60,325 mm (2,375")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk4,1 bar (60 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4,1 bar (60 psi) - MFLX95680-20Transfer Tubing, Ester-Based Polyurethane, 5/16" ID x 7/16" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 5/16" ID x 7/16" OD; 100 Ft8 mm (0,3125")1,6 mm1,2 mm (0,4375")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod2.880,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-20Model95680-20Bøjningsradius49,2125 mm (1,9375")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk4,5 bar (65 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4,5 bar (65 psi) - MFLX95680-28Transfer Tubing, Ester-Based Polyurethane, 7/16" ID x 5/8" OD; 100 FtTransfer Tubing, Ester-Based Polyurethane, 7/16" ID x 5/8" OD; 100 Ft11,2 mm (0,4375")2,4 mm16 mm (0,625")Ester-based PU30,5 mufarvetLog ind for tilgængelighed1 * 100 Fod4.340,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX95680-28Model95680-28Bøjningsradius42,8625 mm (1,6875")ClarityTranslucentLength per pack30,5 m (100 ft.)Maks. tryk5,1 bar (74 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperatur–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5,1 bar (74 psi)