Orders LinkContactUsLinkComponent
S.O.C medium, Corning®
S.O.C medium, Corning®
S.O.C medium, 10
Katalog # MDTC46-003-CR
Leverandør:   Corning
S.O.C medium, Corning®
Katalog # MDTC46-003-CR
Leverandør:  Corning
CAS nummer:  
S.O.C medium, Corning®
S.O.C medium, Corning®
S.O.C medium, 10
CAS nummer:  
Price
909,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
S.O.C
The use of SOC medium improves transformation efficiency.
medium is used as a recovery medium during the final stage of transformation of E. coli.
Specifikationer
Specifikationer
  • Pakn:
    10
  • Beskrivelse:
    S.O.C medium
Specifikationer