Om denne vare
Use under full vacuum.
- Chemical-resistant for aggressive fluids
- Voltage tolerance is ±10%
- 100% duty cycle
Two-way direct lift valves are normally closed and feature bellows made of PTFE and seals made of Viton®.
Choose from valves with a PVC, polypropylene, or PVDF body for a wide range of agreesive fluid applications.
- Environmentally Preferable:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.