Orders LinkContactUsLinkComponent
Pump Motor Spider Inserts, Buna-N®
Pump Motor Spider Inserts, Buna-N®
Spider Insert, Buna-N, for A/L090/L095 Couplings
Katalog # MFLX07127-69
Leverandør:   Avantor Fluid Handling
Pump Motor Spider Inserts, Buna-N®
Katalog # MFLX07127-69
Leverandør:  Avantor Fluid Handling
Pump Motor Spider Inserts, Buna-N®
Pump Motor Spider Inserts, Buna-N®
Spider Insert, Buna-N, for A/L090/L095 Couplings
Price
121,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Create your own motor mounting with the hardware below.
  • Constructed of durable Buna-N®
Spider smooths power transfer, absorbs vibration, and compensates for minor misalignment.
Specifikationer
  • Beskrivelse:
    Spider Insert, Buna-N, for A/L090/L095 Couplings
Accessories
Produktinformation
  • Katalog #
    Motor hp
    Tilgængelighed
    Price
  • Coupling hardware for 5/8" diameter bore, 7,2 hp maximum
    Coupling hardware for 5/8" diameter bore, 7,2 hp maximum
    MFLX07127-59
    7,2
    Log ind for tilgængelighed
    1 * 1 Items - 198,00 kr