Feature micrometer flow adjustment—deliver exact quantities of liquid with minimal pulsation.
- Withstand up to 5000 psi outlet pressure
- Dual-piston models have independent piston control—pump a single liquid, or pump two different liquids at a proportional rate
- Brushless synchronous motors
Inlet accepts 1/8" OD tubing; outlet accepts 1/16" OD high-pressure SS tubing (order separately). Decrease pulsation with pulse dampener model 07115-55.
What's included: All models come with a sturdy aluminum housing and a 7-ft, three-wire cord (115 VAC models have a plug; 220 VAC models have a European plug).
FLUOROLOY G® The Fluorocarbon Co.
KEL-F® 3M Co.
- Environmentally Preferable:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.