ContactUsLinkComponent
Flux® Heavy-Duty Drum Pump Motors, TEFC
Flux F458-1 120V Electric Drum Pump Motor, 3/4 HP, TEFC; 120 VAC
Catalog # MFLX07076-20
CE Compliant
undefined
Flux F458-1 120V Electric Drum Pump Motor, 3/4 HP, TEFC; 120 VAC
Catalog # MFLX07076-20
CE Compliant
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Power:
    120 VAC; 60 Hz
  • Maks. viskositet:
    800 cP
  • Beskrivelse:
    Electric Drum Pump Motor, 3/4 HP, TEFC; 120 VAC
  • CE regulatory approval:
    Yes
  • Motor hp:
    3/4
  • Motor type:
    TEFC
  • Garanti:
    3 Months
  • Pakn:
    1
  • Max. motor RPM:
    9500 rpm
Specifications
About this item
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp
Heavy-duty motors are best for applications requiring high output head. Motors have a built-in overload switch with manual rest to avoid the danger of automatic restarting.
Order tube set and accessories separately.
  • Power:
    120 VAC; 60 Hz
  • Maks. viskositet:
    800 cP
  • Beskrivelse:
    Electric Drum Pump Motor, 3/4 HP, TEFC; 120 VAC
  • Environmentally Preferable:
  • CE regulatory approval:
    Yes
  • Motor hp:
    3/4
  • Motor type:
    TEFC
  • Garanti:
    3 Months
  • Pakn:
    1
  • Max. motor RPM:
    9500 rpm