Orders LinkContactUsLinkComponent
Masterflex® Liquid Detector Pad Cables, Avantor®
Masterflex® Liquid Detector Pad Cables, Avantor®
Replacement RJ-12 cable, 3,5 ft.
Katalog # MFLX77095-02
Leverandør:   Avantor Fluid Handling
Masterflex® Liquid Detector Pad Cables, Avantor®
Katalog # MFLX77095-02
Leverandør:  Avantor Fluid Handling
Masterflex® Liquid Detector Pad Cables, Avantor®
Masterflex® Liquid Detector Pad Cables, Avantor®
Replacement RJ-12 cable, 3,5 ft.
Price
593,00 kr
Log ind for tilgængelighed
1 * 1,067 m
Product Details & Documents
Make the most of your Liqui-Sense leak detector with suitable replacement parts.
  • Compatible with Masterflex® Liqui-Sense controllers
This replacement cable features a 6-pin cable with RJ-12 connections and has a 3,5 foot cable length.
Specifikationer
  • Beskrivelse:
    Replacement RJ-12 cable
  • Længde:
    3,5 ft.
  • Cat. No.:
    MFLX77095-02
  • Model:
    77095-02
Accessories
Produktinformation
  • Katalog #
    Længde
    Tilgængelighed
    Price
  • Replacement RJ-12 cable
    Replacement RJ-12 cable
    MFLX77095-02
    3,5 ft.
    Log ind for tilgængelighed
    1 * 1,067 m - 593,00 kr