Orders LinkContactUsLinkComponent
MultiRack reloads, Axygen®
MultiRack reloads, Axygen®
Supplier:   Corning
MultiRack reloads, Axygen®
Supplier:  Corning
Certificates
MultiRack reloads, Axygen®
MultiRack reloads, Axygen®
4 Options Available Below
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack reload system is designed to fit Axygen® MultiRack as well as racks from other suppliers without tedious realignment. This design not only saves lab space but also uses less plastic to reduce environmental impact. Axygen® MultiRack reload tips are available in a wide range of sizes (10 µl extended, 200, 300 and 1000 µl extended).
Product Family Options
Product Information
  • Spidstype
    Volumen
    Pakket
    Availability
    Price
  • Extended length
    0,1 - 10 µl
    Reload: 960 Tips/Pack, 4 Packs/Case
    Log in for availability
    Log in for availability
    1 * 3.840 Items
    1 * 3.840 Items - 1.310,00 kr
    1.310,00 kr
    1 * 3.840 Items
       1.310,00 kr
  • Standard length
    1 - 200 µl
    Reload: 960 Tips/Pack, 4 Packs/Case
    Log in for availability
    Log in for availability
    1 * 3.840 Items
    1 * 3.840 Items - 1.310,00 kr
    1.310,00 kr
    1 * 3.840 Items
       1.310,00 kr
  • Standard length
    1 - 300 µl
    Reload: 960 Tips/Pack, 4 Packs/Case
    Log in for availability
    Log in for availability
    1 * 3.840 Items
    1 * 3.840 Items - 1.350,00 kr
    1.350,00 kr
    1 * 3.840 Items
       1.350,00 kr
  • Extended length
    50 - 1250 µl
    Reload: 768 Tips/Pack, 4 Packs/Case
    Log in for availability
    Log in for availability
    1 * 3.072 Items
    1 * 3.072 Items - 1.310,00 kr
    1.310,00 kr
    1 * 3.072 Items
       1.310,00 kr