Orders LinkContactUsLinkComponent
Dollies
Dollies
Leverandør:   TEKNOMEK
Dollies
Leverandør:  TEKNOMEK
Certifikater
Dollies
7 tilgængelige muligheder nedenfor
Product Details & Documents
Our stainless steel circular dollies are a extremely robust, with an up to 150 kg payload.
  • Fully welded 304 grade stainless steel
  • Brushed dull polish finish
  • 4 swivel castors
  • Crevice free
  • Sturdy design
  • Easy-clean design
  • Fully welded, ready to use
They come with swivel castors, ensuring smooth and effortless movement. Rectangular stainless steel and aluminum bogies are also available.
Product Family Options
Circular
Produktinformation
  • B×D×H
    Tilgængelighed
    Price
  • TEKMBS0010/300
    Circular dollie, 300 mm Ø stainless steel
    Circular dollie, 300 mm Ø stainless steel
    300×300×165 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • TEKMBS0010/400
    Circular dollie, 400 mm Ø stainless steel
    Circular dollie, 400 mm Ø stainless steel
    400×400×165 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • TEKMBS0010/450
    Circular dollie, 450 mm Ø stainless steel
    Circular dollie, 450 mm Ø stainless steel
    450×450×165 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • TEKMBS0010/460
    Circular dollie, 460 mm Ø stainless steel
    Circular dollie, 460 mm Ø stainless steel
    460×460×165 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • TEKMBS0010/600
    Circular dollie, 600 mm Ø stainless steel
    Circular dollie, 600 mm Ø stainless steel
    600×600×165 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Rectangular
Produktinformation
  • B×D×H
    Tilgængelighed
    Price
  • TEKMBS100006040S
    Mobile roller bogie, stainless steel for 600×400 mm tray
    Mobile roller bogie, stainless steel for 600×400 mm tray
    600×400×175 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • TEKMBS100007545S
    Mobile roller bogie stainless steel for 750×450 mm tray
    Mobile roller bogie stainless steel for 750×450 mm tray
    750×450×175 mm