Orders LinkContactUsLinkComponent
VWR®, Snap Ring Vials ND11
VWR®, Snap Ring Vials ND11
VWR®, Snap Ring Vials ND11
Leverandør:  VWR Collection
Certifikater
VWR®, Snap Ring Vials ND11
VWR®, Snap Ring Vials ND11
30 tilgængelige muligheder nedenfor
Product Details & Documents
Snap ring ND11 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyethylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Wide opening
Snap ring ND11 vials constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear version with a flat bottom to ensure compatibility with inserts. Multiple cap colours mostly available with assembled septa are engineered to prevent sample contamination while providing consistent, secure seals. Furthermore, snap ring vials ND11 are available in amber and transparent plastic version.
Product Family Options
Vials
Produktinformation
  • Kapacitet
    Grundlag
    Diameter×H
    Materiale
    Farve
    Kasket inkluderet
    Låg type
    Cap størrelse
    Hættemateriale
    Tilgængelighed
    Price
  • 548-0429A
    Micro-vials with integrated micro-glass insert
    Micro-vials with integrated micro-glass insert
    0,20 ml
    Conical bottom, self-standing
    11,6×32 mm
    TPX
    transparent
    No
    Crimp/snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 8.140,00 kr
    8.140,00 kr
    1 * 1.000 Items
       8.140,00 kr
  • 548-0120A
    Micro-vials, high purity
    Micro-vials, high purity
    0,30 ml
    Conical bottom, self-standing
    11,6×32 mm
    PP
    transparent
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.560,00 kr
    2.560,00 kr
    1 * 1.000 Items
       2.560,00 kr
  • 548-0451A
    Micro-vials
    Micro-vials
    0,30 ml
    Conical bottom, self-standing
    11,6×32 mm
    TPX
    transparent
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.670,00 kr
    3.670,00 kr
    1 * 1.000 Items
       3.670,00 kr
  • 548-0454A
    Micro-vials
    Micro-vials
    0,30 ml
    Conical bottom, self-standing
    11,6×32 mm
    PP
    ravfarvet
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.710,00 kr
    2.710,00 kr
    1 * 1.000 Items
       2.710,00 kr
  • 548-1441A
    Vials with integrated micro-insert, base bonded
    Vials with integrated micro-insert, base bonded
    0,30 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Crimp/snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 4.980,00 kr
    4.980,00 kr
    1 * 1.000 Items
       4.980,00 kr
  • 548-1472A
    Vials with integrated micro-insert
    Vials with integrated micro-insert
    0,30 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    ravfarvet
    No
    Crimp/snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 5.340,00 kr
    5.340,00 kr
    1 * 1.000 Items
       5.340,00 kr
  • 548-0895A
    Micro-vials
    Micro-vials
    0,70 ml
    Flat bottom
    11,6×32 mm
    PP
    transparent
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.930,00 kr
    1.930,00 kr
    1 * 1.000 Items
       1.930,00 kr
  • Vials
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.830,00 kr
    1.830,00 kr
    1 * 1.000 Items
       1.830,00 kr
  • 548-0012A
    Vials with write-on patch
    Vials with write-on patch
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    ravfarvet
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1.000 Items
       1.820,00 kr
  • 548-0422A
    Vials with write-on patch
    Vials with write-on patch
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1.000 Items
       1.820,00 kr
  • 548-1859A
    Vials with write-on patch, silanized
    Vials with write-on patch, silanized
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    ravfarvet
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.720,00 kr
    3.720,00 kr
    1 * 1.000 Items
       3.720,00 kr
  • 548-1860A
    Vials, silanized
    Vials, silanized
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.620,00 kr
    3.620,00 kr
    1 * 1.000 Items
       3.620,00 kr
  • Vials
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 190,00 kr
    190,00 kr
    1 * 100 Items
       190,00 kr
  • 548-8015A
    Vials with write-on patch
    Vials with write-on patch
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 196,00 kr
    196,00 kr
    1 * 100 Items
       196,00 kr
  • 548-8016A
    Vials with write-on patch
    Vials with write-on patch
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    ravfarvet
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 218,00 kr
    218,00 kr
    1 * 100 Items
       218,00 kr
  • Vials
    0,90 ml
    Conical bottom (maximum recovery)
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 23.730,00 kr
    23.730,00 kr
    1 * 1.000 Items
       23.730,00 kr
  • 548-2192A
    Micro-vials
    Micro-vials
    0,70 ml
    Flat bottom
    11,6×32 mm
    PP
    transparent
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.010,00 kr
    2.010,00 kr
    1 * 1.000 Items
       2.010,00 kr
  • Vials
    1,10 ml
    Solid glass bottom, round, conical
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 7.160,00 kr
    7.160,00 kr
    1 * 1.000 Items
       7.160,00 kr
  • Vials
    1,10 ml
    Solid glass bottom, round, conical
    11,6×32 mm
    Borosilicate glass 5.1
    ravfarvet
    No
    Snap
    ND11
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 7.840,00 kr
    7.840,00 kr
    1 * 1.000 Items
       7.840,00 kr
Vial kits
Produktinformation
  • Kapacitet
    Grundlag
    Diameter×H
    Materiale
    Farve
    Kasket inkluderet
    Låg type
    Cap størrelse
    Hættemateriale
    Tilgængelighed
    Price
  • 548-2368A
    Vial kits with write-on patch
    Vial kits with write-on patch
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.170,00 kr
    3.170,00 kr
    1 * 1.000 Items
       3.170,00 kr
  • 548-2367A
    Vial kits with write-on patch, high purity
    Vial kits with write-on patch, high purity
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    ravfarvet
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.900,00 kr
    3.900,00 kr
    1 * 1.000 Items
       3.900,00 kr
  • 548-2366A
    Vial kits with write-on patch, high purity
    Vial kits with write-on patch, high purity
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.880,00 kr
    3.880,00 kr
    1 * 1.000 Items
       3.880,00 kr
  • 548-0471A
    Vial kits, high purity
    Vial kits, high purity
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 409,00 kr
    409,00 kr
    1 * 100 Items
       409,00 kr
  • 548-1956A
    Vial kits with write-on patch
    Vial kits with write-on patch
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 4.400,00 kr
    4.400,00 kr
    1 * 1.000 Items
       4.400,00 kr
  • 548-0472A
    Vial kits, high purity
    Vial kits, high purity
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 518,00 kr
    518,00 kr
    1 * 100 Items
       518,00 kr
  • 548-0470A
    Vial kits
    Vial kits
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 340,00 kr
    340,00 kr
    1 * 100 Items
       340,00 kr
  • 548-0473A
    Vial kits
    Vial kits
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 512,00 kr
    512,00 kr
    1 * 100 Items
       512,00 kr
  • 548-1930A
    Vial kits, approved IM quality
    Vial kits, approved IM quality
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 371,00 kr
    371,00 kr
    1 * 100 Items
       371,00 kr
Vial kits, assembled
Produktinformation
  • Kapacitet
    Grundlag
    Diameter×H
    Materiale
    Farve
    Kasket inkluderet
    Låg type
    Cap størrelse
    Hættemateriale
    Tilgængelighed
    Price
  • 548-2175A
    Vial kits with write-on patch
    Vial kits with write-on patch
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    ravfarvet
    Yes
    Crimp
    ND11
    Aluminium
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.120,00 kr
    3.120,00 kr
    1 * 1.000 Items
       3.120,00 kr
  • 548-3285A
    Vial kits
    Vial kits
    1,50 ml
    Flat bottom
    11,6×32 mm
    Borosilicate glass 5.1
    klar
    Yes
    Snap
    ND11
    PP
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 5.020,00 kr
    5.020,00 kr
    1 * 1.000 Items
       5.020,00 kr
Snap caps, open top
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    53° shore D
    Snap caps, open top
    blå
    PTFE virginal
    0,2 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.970,00 kr
    2.970,00 kr
    1 * 1.000 Items
       2.970,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    transparent
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.530,00 kr
    1.530,00 kr
    1 * 1.000 Items
       1.530,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    transparent
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    gul
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.520,00 kr
    2.520,00 kr
    1 * 1.000 Items
       2.520,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, approved IM quality
    transparent
    Red rubber/beige PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.690,00 kr
    1.690,00 kr
    1 * 1.000 Items
       1.690,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    transparent
    White silicone/red PTFE, Y-slit
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.690,00 kr
    3.690,00 kr
    1 * 1.000 Items
       3.690,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    transparent
    White silicone/red PTFE, Y-slit
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.690,00 kr
    3.690,00 kr
    1 * 1.000 Items
       3.690,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    transparent
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.530,00 kr
    1.530,00 kr
    1 * 1.000 Items
       1.530,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    transparent
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    transparent
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.180,00 kr
    2.180,00 kr
    1 * 1.000 Items
       2.180,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    grøn
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.740,00 kr
    1.740,00 kr
    1 * 1.000 Items
       1.740,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top, high purity
    transparent
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.160,00 kr
    3.160,00 kr
    1 * 1.000 Items
       3.160,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    gul
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.740,00 kr
    1.740,00 kr
    1 * 1.000 Items
       1.740,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top
    transparent
    White silicone/blue PTFE, Y-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.790,00 kr
    3.790,00 kr
    1 * 1.000 Items
       3.790,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    blå
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.520,00 kr
    2.520,00 kr
    1 * 1.000 Items
       2.520,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    blå
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.740,00 kr
    1.740,00 kr
    1 * 1.000 Items
       1.740,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    blå
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top
    blå
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.900,00 kr
    3.900,00 kr
    1 * 1.000 Items
       3.900,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    rød
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.740,00 kr
    1.740,00 kr
    1 * 1.000 Items
       1.740,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    rød
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    rød
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.520,00 kr
    2.520,00 kr
    1 * 1.000 Items
       2.520,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top
    rød
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.900,00 kr
    3.900,00 kr
    1 * 1.000 Items
       3.900,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    gul
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top
    gul
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.900,00 kr
    3.900,00 kr
    1 * 1.000 Items
       3.900,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, approved IM quality
    blå
    Red rubber/beige PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1.000 Items
       1.820,00 kr
Produktinformation
  • Katalog #
    Materiale
    Lågfarve
    Beskrivelse
    Cap størrelse
    Septum
    Tykkelse
    Hårdhed
    Tilgængelighed
    Price
  • ND11
    transparent
    Snap caps, open top
    PTFE virginal
    28
    0,2 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.270,00 kr
    1.270,00 kr
    1 * 1.000 Items
       1.270,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    lyserød
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.180,00 kr
    2.180,00 kr
    1 * 1.000 Items
       2.180,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    lyserød
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    lyserød
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.160,00 kr
    3.160,00 kr
    1 * 1.000 Items
       3.160,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    grøn
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.520,00 kr
    2.520,00 kr
    1 * 1.000 Items
       2.520,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    transparent
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.180,00 kr
    2.180,00 kr
    1 * 1.000 Items
       2.180,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    blå
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.740,00 kr
    1.740,00 kr
    1 * 1.000 Items
       1.740,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top
    transparent
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.510,00 kr
    3.510,00 kr
    1 * 1.000 Items
       3.510,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, high purity
    blå
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.520,00 kr
    2.520,00 kr
    1 * 1.000 Items
       2.520,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top
    blå
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.900,00 kr
    3.900,00 kr
    1 * 1.000 Items
       3.900,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    blå
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    blå
    White silicone/red PTFE, Y-slit
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 4.140,00 kr
    4.140,00 kr
    1 * 1.000 Items
       4.140,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    blå
    White silicone/red PTFE, Y-slit
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 4.140,00 kr
    4.140,00 kr
    1 * 1.000 Items
       4.140,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    grøn
    Red PTFE/white silicone/red PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1.000 Items
       3.100,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    60° shore A
    Snap caps, open top
    transparent
    Red-orange natural rubber/transparent TEF
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 185,00 kr
    185,00 kr
    1 * 100 Items
       185,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, approved IM quality
    transparent
    Red rubber/beige PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.690,00 kr
    1.690,00 kr
    1 * 1.000 Items
       1.690,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, approved IM quality
    blå
    Red rubber/beige PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1.000 Items
       1.820,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    transparent
    Red rubber/beige PTFE
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 208,00 kr
    208,00 kr
    1 * 100 Items
       208,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top
    transparent
    White silicone/red PTFE
    1,3 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 283,00 kr
    283,00 kr
    1 * 100 Items
       283,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    55° shore A
    Snap caps, open top
    transparent
    White silicone/blue PTFE, cross-slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 421,00 kr
    421,00 kr
    1 * 100 Items
       421,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, Ecoline
    blå
    White silicone/beige PTFE, cross-slitted
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2.000 Items
    1 * 2.000 Items - 2.940,00 kr
    2.940,00 kr
    1 * 2.000 Items
       2.940,00 kr
Produktinformation
  • Katalog #
    Cap størrelse
    Hårdhed
    Beskrivelse
    Lågfarve
    Septum
    Tykkelse
    Materiale
    Tilgængelighed
    Price
  • ND11
    45° shore A
    Snap caps, open top, Ecoline
    blå
    White silicone/beige PTFE, slit
    1,0 mm
    PE
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2.000 Items
    1 * 2.000 Items - 7.440,00 kr
    7.440,00 kr
    1 * 2.000 Items
       7.440,00 kr
Septa ND11
Produktinformation
  • Katalog #
    Septum
    Hårdhed
    Tykkelse
    Tilgængelighed
    Price
  • White silicone/red PTFE
    45° shore A
    1,3 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.410,00 kr
    2.410,00 kr
    1 * 1.000 Items
       2.410,00 kr
Produktinformation
  • Katalog #
    Septum
    Hårdhed
    Tykkelse
    Tilgængelighed
    Price
  • PTFE virginal
    53° shore D
    0,25 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 682,00 kr
    682,00 kr
    1 * 1.000 Items
       682,00 kr
Produktinformation
  • Katalog #
    Septum
    Hårdhed
    Tykkelse
    Tilgængelighed
    Price
  • Red PTFE/white silicone/red PTFE
    45° shore A
    1,0 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.940,00 kr
    2.940,00 kr
    1 * 1.000 Items
       2.940,00 kr
Produktinformation
  • Katalog #
    Septum
    Hårdhed
    Tykkelse
    Tilgængelighed
    Price
  • Viton 1A black
    70° shore A
    1,5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 5.240,00 kr
    5.240,00 kr
    1 * 1.000 Items
       5.240,00 kr
Produktinformation
  • Katalog #
    Septum
    Hårdhed
    Tykkelse
    Tilgængelighed
    Price
  • White silicone/blue PTFE, cross-slit
    55° shore A
    1,5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 2.850,00 kr
    2.850,00 kr
    1 * 1.000 Items
       2.850,00 kr
Produktinformation
  • Katalog #
    Septum
    Hårdhed
    Tykkelse
    Tilgængelighed
    Price
  • Red rubber/beige PTFE
    45° shore A
    1,0 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.610,00 kr
    1.610,00 kr
    1 * 1.000 Items
       1.610,00 kr
Produktinformation
  • Katalog #
    Septum
    Hårdhed
    Tykkelse
    Tilgængelighed
    Price
  • Red-orange natural rubber/transparent TEF
    60° shore A
    1,0 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 948,00 kr
    948,00 kr
    1 * 1.000 Items
       948,00 kr