ContactUsLinkComponent
Escherichia coli WDCM 00012 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Escherichia coli WDCM 00012 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000127-10EA
Supplier:  Merck
CAS Number:  
Escherichia coli WDCM 00012 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000127-10EA
Supplier:  Merck
CAS Number:  
Specifications
  • Pakn:
    10
  • Beskrivelse:
    Escherichia coli WDCM 00012 Vitroids™, 50000 - 150000 CFU mean value range
  • M-Clarity quality level:
    MQ300
  • Opbevaringstemperatur:
    –20 °C
  • Product type:
    Hydrosoluble discs
Specifications
About this item
Vitroids™ discs contain Escherichia coli WDCM 00012 viable microorganisms in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.
  • Defined CFU range and low standard deviation
  • Fast, reliable and easy to use
The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year and are in a viable state with a shelf life of 1-3 years.
Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.
Vitroids™ are derived from a traceable culture obtained freeze-dried from CECT®.
Certifications: These certified reference materials (CRMs) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025.