Orders LinkContactUsLinkComponent
Flexible cable conduit, b.safe
Flexible cable conduit, b.safe
Flexible cable conduit, 10 m length
Catalog # BOHLM564-01
Supplier:   Bohlender
Flexible cable conduit, b.safe
Catalog # BOHLM564-01
Supplier:  Bohlender
Flexible cable conduit, b.safe
Flexible cable conduit, b.safe
Flexible cable conduit, 10 m length
Price
358,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Expandable cable conduit to bundle several hoses or signal cables, can be cut to the required length.
  • Made of PE
  • Bundle several hoses or signal cables
  • Can be cut to size
Save space by bundling parallel hose lines or signal cables.
Specifications
  • Beskrivelse:
    Flexible cable conduit, 10 m length