Impact goggle, Martcare®
Martcare® impact goggle, Polycarbonate
Catalog # JSPAAGC010-301-300
Supplier: JSP
Jump to:
Product Details & Documents
A cost-effective, compact, and lightweight goggle.
- Compact and lightweight
- Tough polycarbonate lens
- Impact protection
- Vented body
- PVC frame
A compact and lightweight goggle with tough polycarbonate lens and PVC frame, offering high resistance to impact from flying particles.
Certifications: Conforms to EN 166.1.B.
Documents
Specifications
- Beskrivelse:Martcare® impact goggle
- Linser:Polycarbonate