Orders LinkContactUsLinkComponent
Sampling head, MD8 Airscan®
Sampling head, MD8 Airscan®
MD8 air samplers stainless steel sampling Head Bayonet, stainless steel with silicone seal
Katalog # 514-1293
Leverandør:   Sartorius
Sampling head, MD8 Airscan®
Katalog # 514-1293
Leverandør:  Sartorius
Sampling head, MD8 Airscan®
MD8 air samplers stainless steel sampling Head Bayonet, stainless steel with silicone seal
Price
53.810,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Sampling head with bayonet connection.
  • Flexible operating conditions
  • Generates a vacuum and pumps a defined air sample volume from the ambient air
The bayonet connector accommodates the filter adapter. Sampling heads with a bayonet connection are particularly suitable for applications where the sampling head is not installed in a system.
Bestillingsoplysninger: Supplied without accessories.
Specifikationer
  • Beskrivelse:
    MD8 air samplers stainless steel sampling Head Bayonet, stainless steel with silicone seal