Stopcock, b.safe
10 mm, GL 25 (Male and female)
BOHLR1203-01
The grip position indicates if the passage is open or closed.
- Made of static dissipative PE-EX
- Firmly mounted union nut made of PPS-EX with female thread GL 25
- Integrated stopcock to interrupt flow
- Outer dimension: 42×55×116 mm
Connecting piece made of static dissipative PE-EX with male GL 25 thread, permanently fixed union nut made of PPS-EX with female GL 25 thread and integrated stopcock to interrupt flow. The grip position indicates if the passage is open or closed.
- Hul Ø:10 mm
- Gevind:GL 25 (Male and female)