Colour Lab Tapes
Om denne vare
Colour laboratory tape is available in a wide range of colours and widths, for easy identification and colour-coding.
- Temperature range: –80 to +121 °C
- Ideal for all general-use applications in the lab
- Continuous tape format allows use of as much or little as necessary
- Resistant to gamma irradiation (tested to up to 50 kGy)
- Available in a wide range of colors and widths, for easy identification and colour-coding
Available in an assortment of eye-catching colours and various widths. Lab tape is perfect for colour-coding and general lab use. Can be written on with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
30 tilgængelige muligheder nedenfor
Product Details & Documents
Colour laboratory tape is available in a wide range of colours and widths, for easy identification and colour-coding.
- Temperature range: –80 to +121 °C
- Ideal for all general-use applications in the lab
- Continuous tape format allows use of as much or little as necessary
- Resistant to gamma irradiation (tested to up to 50 kGy)
- Available in a wide range of colors and widths, for easy identification and colour-coding
Available in an assortment of eye-catching colours and various widths. Lab tape is perfect for colour-coding and general lab use. Can be written on with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Product Family Options
Produktinformation
- FarveLængdeBreddeTilgængelighedPrice
- GAINPAT-13GAColour lab tape, core size: 76,2 mm
- GAINPAT-13GRColour lab tape, core size: 76,2 mm
- GAINPAT-13GSColour lab tape, core size: 76,2 mm
- GAINPAT-13LBLColour lab tape, core size: 76,2 mm
- GAINPAT-13ORColour lab tape, core size: 76,2 mm
- GAINPAT-13PIColour lab tape, core size: 76,2 mm
- GAINPAT-13PUColour lab tape, core size: 76,2 mm
- GAINPAT-13REColour lab tape, core size: 76,2 mm
- GAINPAT-13WHColour lab tape, core size: 76,2 mm
- GAINPAT-13YEColour lab tape, core size: 76,2 mm
- GAINPAT-18GAColour lab tape, core size: 76,2 mm
- GAINPAT-18GRColour lab tape, core size: 76,2 mm
- GAINPAT-18GSColour lab tape, core size: 76,2 mm
- GAINPAT-18LBLColour lab tape, core size: 76,2 mm
- GAINPAT-18ORColour lab tape, core size: 76,2 mm
- GAINPAT-18PIColour lab tape, core size: 76,2 mm
- GAINPAT-18PUColour lab tape, core size: 76,2 mm
- GAINPAT-18REColour lab tape, core size: 76,2 mm
- GAINPAT-18WHColour lab tape, core size: 76,2 mm
- GAINPAT-18YEColour lab tape, core size: 76,2 mm
- GAINPAT-24GAColour lab tape, core size: 76,2 mm
- GAINPAT-24GRColour lab tape, core size: 76,2 mm
- GAINPAT-24GSColour lab tape, core size: 76,2 mm
- GAINPAT-24LBLColour lab tape, core size: 76,2 mm
- GAINPAT-24ORColour lab tape, core size: 76,2 mm
- GAINPAT-24PIColour lab tape, core size: 76,2 mm
- GAINPAT-24PUColour lab tape, core size: 76,2 mm
- GAINPAT-24REColour lab tape, core size: 76,2 mm
- GAINPAT-24WHColour lab tape, core size: 76,2 mm
- GAINPAT-24YEColour lab tape, core size: 76,2 mm