Orders LinkContactUsLinkComponent
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
Leverandør:   GA
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
Leverandør:  GA
Certifikater
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
109 tilgængelige muligheder nedenfor
Product Details & Documents
These labels are available in a range of sizes and configurations, perfect for identifying small diameter cryo vials, microtubes, cardboard or polypropylene cryo/freezer boxes, cell culture plates, and plastic bags.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Resistant to long term cryogenic storage in freezers and liquid nitrogen (−196 °C/−321 °F)
  • Resistant to spraying and swabbing with alcohol and surface sanitising wipes
  • Require no cure time before liquid nitrogen immersion; will also adhere to wet surfaces
  • High-quality thermal transfer printing, ideal for serialised data and barcodes
  • Available in a large range of sizes, colours, and configurations
  • RR-class resin ribbon recommended
These thermal transfer cryogenic labels have a strong permanent adhesive that will remain firmly and permanently attached in ultra-low temperatures. They are designed for long-term cryo-preservation under cryogenic laboratory conditions, including in liquid and vapour phase liquid nitrogen (−196 °C/−321 °F), lab freezers (−80 °C/−112 °F), and dry ice transportation. JTTA labels require zero cure time and can be immersed in liquid nitrogen immediately after application without the need for additional tape or lamination. Additionally, these labels are waterproof, weatherproof, and durable. JTTA labels will adhere to wet surfaces, resist multiple freeze-thaw cycles, and can withstand elevated temperatures up to 110 °C.
They are also resistant to spraying with ethanol, isopropanol, and other alcohols, bleach, and detergents (testing recommended), as well as routine wiping and rubbing with surface sanitizing wipes such as Sani- Wipe™, Eco-Wipe™, and Eco-Wipe™ Duo.
These labels can be printed with all major brands of thermal-transfer printers. They are available in a range of colours and sizes and can be printed with alphanumeric text, as well as 1D and 2D barcodes.
Product Family Options
Produktinformation
  • Farve
    Dimensioner
    Pakket
    Tilgængelighed
    Price
  • GAINJTTA-204C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    11,9×11,9 mm (0,47×0,47")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 577,00 kr
    577,00 kr
    1 * 1 ROL
       577,00 kr
  • GAINJTTA-204C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    11,9×11,9 mm (0,47×0,47")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.160,00 kr
    1.160,00 kr
    1 * 1 ROL
       1.160,00 kr
  • GAINJTTA-162C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    12,7×25,4 mm (0,5×1")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 721,00 kr
    721,00 kr
    1 * 1 ROL
       721,00 kr
  • GAINJTTA-277C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    15,9×30,5 mm (0,63×1,2")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 356,00 kr
    356,00 kr
    1 * 1 ROL
       356,00 kr
  • GAINJTTA-277C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    15,9×30,5 mm (0,63×1,2")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.070,00 kr
    1.070,00 kr
    1 * 1 ROL
       1.070,00 kr
  • GAINJTTA51NPC1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    19,1×10,2 mm (0,75×0,4")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 721,00 kr
    721,00 kr
    1 * 1 ROL
       721,00 kr
  • GAINJTTA-49C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    19,1×15,9 mm (0,75×0,63")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 697,00 kr
    697,00 kr
    1 * 1 ROL
       697,00 kr
  • GAINJTTA-49C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    19,1×15,9 mm (0,75×0,63")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 697,00 kr
    697,00 kr
    1 * 1 ROL
       697,00 kr
  • GAINJTTA-69C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    19,1×19,1 mm (0,75×0,75")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 806,00 kr
    806,00 kr
    1 * 1 ROL
       806,00 kr
  • GAINJTTA-69C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    19,1×19,1 mm (0,75×0,75")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 806,00 kr
    806,00 kr
    1 * 1 ROL
       806,00 kr
  • GAINJTTA-177C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    19,1×38,1 mm (0,75×1,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 521,00 kr
    521,00 kr
    1 * 1 ROL
       521,00 kr
  • GAINJTTA213NPC15WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    19,1×5,1 mm (0,75×0,2")
    5000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.450,00 kr
    1.450,00 kr
    1 * 1 ROL
       1.450,00 kr
  • GAINJTTA-98C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    22,2×22,2 mm (0,87×0,87")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 330,00 kr
    330,00 kr
    1 * 1 ROL
       330,00 kr
  • GAINJTTA-98C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    22,2×22,2 mm (0,87×0,87")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 989,00 kr
    989,00 kr
    1 * 1 ROL
       989,00 kr
  • GAINJTTA188NPC13WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    22,9×12,7 mm (0,9×0,5")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 865,00 kr
    865,00 kr
    1 * 1 ROL
       865,00 kr
  • GAINJTTA-117C1-3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    22×15 mm (0,87×0,59")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 865,00 kr
    865,00 kr
    1 * 1 ROL
       865,00 kr
  • GAINJTTA-117C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    22×15 mm (0,87×0,59")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
  • GAINJTTA-67C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    23,8×23,8 mm (0,94×0,94")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
  • GAINJTTA-67C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    23,8×23,8 mm (0,94×0,94")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
  • GAINJTTA-152C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    23×19,1 mm (0,9×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 324,00 kr
    324,00 kr
    1 * 1 ROL
       324,00 kr
  • GAINJTTA-152C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    23×19,1 mm (0,9×0,75")
    6000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.940,00 kr
    1.940,00 kr
    1 * 1 ROL
       1.940,00 kr
  • GAINJTTA-1C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4 × 11,1 mm (1×0,44")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 289,00 kr
    289,00 kr
    1 * 1 ROL
       289,00 kr
  • GAINJTTA-1C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    25,4 × 11,1 mm (1×0,44")
    6000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.730,00 kr
    1.730,00 kr
    1 * 1 ROL
       1.730,00 kr
  • GAINJTTA-7C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×12,7 mm (1×0,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 289,00 kr
    289,00 kr
    1 * 1 ROL
       289,00 kr
  • GAINJTTA-7SBC1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×12,7 mm (1×0,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 289,00 kr
    289,00 kr
    1 * 1 ROL
       289,00 kr
  • GAINJTTA-7C3-5WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    25,4×12,7 mm (1×0,5")
    5000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.450,00 kr
    1.450,00 kr
    1 * 1 ROL
       1.450,00 kr
  • GAINJTTA-4C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×19,1 mm (1×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 333,00 kr
    333,00 kr
    1 * 1 ROL
       333,00 kr
  • GAINJTTA-4C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    25,4×19,1 mm (1×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 333,00 kr
    333,00 kr
    1 * 1 ROL
       333,00 kr
  • GAINJTTA-29C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×25,4 mm (1×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 361,00 kr
    361,00 kr
    1 * 1 ROL
       361,00 kr
  • GAINJTTA-29C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    25,4×25,4 mm (1×1")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.090,00 kr
    1.090,00 kr
    1 * 1 ROL
       1.090,00 kr
  • GAINJTTA-118C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×31,8 mm (1×1,25")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 442,00 kr
    442,00 kr
    1 * 1 ROL
       442,00 kr
  • GAINJTTA-118C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    25,4×31,8 mm (1×1,25")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.770,00 kr
    1.770,00 kr
    1 * 1 ROL
       1.770,00 kr
  • GAINJTTA-94C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×50,8 mm (1×2")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 721,00 kr
    721,00 kr
    1 * 1 ROL
       721,00 kr
  • GAINJTTA209C3-10WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    25,4×6,4 mm (1×0,25")
    10000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 2.840,00 kr
    2.840,00 kr
    1 * 1 ROL
       2.840,00 kr
  • GAINJTTA-209C1-4WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×6,4 mm (1×0,25")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
  • GAINJTTA-225C1-3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    25,4×9,5 mm (1×0,375")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 865,00 kr
    865,00 kr
    1 * 1 ROL
       865,00 kr
  • GAINJTTA-225C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    25,4×9,5 mm (1×0,375")
    6000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.730,00 kr
    1.730,00 kr
    1 * 1 ROL
       1.730,00 kr
  • GAINJTTA-57C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    28,6×19,1 mm (1,12×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
  • GAINJTTA-57C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    28,6×19,1 mm (1,12×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
  • GAINJTTA-57C3-1GA
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    grøn
    28,6×19,1 mm (1,12×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
  • GAINJTTA-18C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    31,8×12,7 mm (1,25×0,5")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 704,00 kr
    704,00 kr
    1 * 1 ROL
       704,00 kr
  • GAINJTTA-18C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    31,8×12,7 mm (1,25×0,5")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 704,00 kr
    704,00 kr
    1 * 1 ROL
       704,00 kr
  • GAINJTTA-9C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    31,8×22,2 mm (1,25×0,875")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 394,00 kr
    394,00 kr
    1 * 1 ROL
       394,00 kr
  • GAINJTTA-9C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    31,8×22,2 mm (1,25×0,875")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 394,00 kr
    394,00 kr
    1 * 1 ROL
       394,00 kr
  • GAINJTTA-58C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    31,8×25,4 mm (1,25×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 442,00 kr
    442,00 kr
    1 * 1 ROL
       442,00 kr
  • GAINJTTA-58C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    31,8×25,4 mm (1,25×1")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.770,00 kr
    1.770,00 kr
    1 * 1 ROL
       1.770,00 kr
  • GAINJTTA-65C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    31,8×31,8 mm (1,25×1,25")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 539,00 kr
    539,00 kr
    1 * 1 ROL
       539,00 kr
  • GAINJTTA-65C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    31,8×31,8 mm (1,25×1,25")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 539,00 kr
    539,00 kr
    1 * 1 ROL
       539,00 kr
  • GAINJTTA-169C1-4WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    31,8×6,4 mm (1,25×0,25")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
  • GAINJTTA-97C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    32,5×15,9 mm (1,28×0,63")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 339,00 kr
    339,00 kr
    1 * 1 ROL
       339,00 kr
  • GAINJTTA-97C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    32,5×15,9 mm (1,28×0,63")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 675,00 kr
    675,00 kr
    1 * 1 ROL
       675,00 kr
  • GAINJTTA-19C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    33×15,9 mm (1,3×0,63")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 339,00 kr
    339,00 kr
    1 * 1 ROL
       339,00 kr
  • GAINJTTA-19C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    33×15,9 mm (1,3×0,63")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 675,00 kr
    675,00 kr
    1 * 1 ROL
       675,00 kr
  • GAINJTTA-6C1-3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    34,9×12,7 mm (1,38×0,5")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 943,00 kr
    943,00 kr
    1 * 1 ROL
       943,00 kr
  • GAINJTTA-168C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    35,6×12,7 mm (1,4×0,5")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 673,00 kr
    673,00 kr
    1 * 1 ROL
       673,00 kr
  • GAINJTTA-168C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    35,6×12,7 mm (1,4×0,5")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.350,00 kr
    1.350,00 kr
    1 * 1 ROL
       1.350,00 kr
  • GAINJTTA-536C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    38,1×10,2 mm (1,5×0,4")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 655,00 kr
    655,00 kr
    1 * 1 ROL
       655,00 kr
  • GAINJTTA-240C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    38,1×12,7 mm (1,5×0,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 341,00 kr
    341,00 kr
    1 * 1 ROL
       341,00 kr
  • GAINJTTA-240C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    38,1×12,7 mm (1,5×0,5")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.020,00 kr
    1.020,00 kr
    1 * 1 ROL
       1.020,00 kr
  • GAINJTTA-10C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    38,1×19,1 mm (1,5×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 406,00 kr
    406,00 kr
    1 * 1 ROL
       406,00 kr
  • GAINJTTA-10C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    38,1×19,1 mm (1,5×0,75")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 810,00 kr
    810,00 kr
    1 * 1 ROL
       810,00 kr
  • GAINJTTA-107C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    38,1×22,2 mm (1,5×0,875")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 463,00 kr
    463,00 kr
    1 * 1 ROL
       463,00 kr
  • GAINJTTA-107C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    38,1×22,2 mm (1,5×0,875")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.390,00 kr
    1.390,00 kr
    1 * 1 ROL
       1.390,00 kr
  • GAINJTTA-504C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    38,1×25,4 mm (1,5×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 545,00 kr
    545,00 kr
    1 * 1 ROL
       545,00 kr
  • GAINJTTA-504C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    38,1×25,4 mm (1,5×1")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.630,00 kr
    1.630,00 kr
    1 * 1 ROL
       1.630,00 kr
  • GAINJTTA-272C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    38,1×31,8 mm (1,5×1,25")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 667,00 kr
    667,00 kr
    1 * 1 ROL
       667,00 kr
  • GAINJTTA-272C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    38,1×31,8 mm (1,5×1,25")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 2.670,00 kr
    2.670,00 kr
    1 * 1 ROL
       2.670,00 kr
  • GAINJTTA-66C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    38,1×38,1 mm (1,5×1,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 751,00 kr
    751,00 kr
    1 * 1 ROL
       751,00 kr
  • GAINJTTA-66C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    38,1×38,1 mm (1,5×1,5")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.510,00 kr
    1.510,00 kr
    1 * 1 ROL
       1.510,00 kr
  • GAINJTTA-232C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    41,3×15,9 mm (1,63×0,63")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 749,00 kr
    749,00 kr
    1 * 1 ROL
       749,00 kr
  • GAINJTTA-232C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    41,3×15,9 mm (1,63×0,63")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.500,00 kr
    1.500,00 kr
    1 * 1 ROL
       1.500,00 kr
  • GAINJTTA-38C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    44,5×12,7 mm (1,75×0,5")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 670,00 kr
    670,00 kr
    1 * 1 ROL
       670,00 kr
  • GAINJTTA-38C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    44,5×12,7 mm (1,75×0,5")
    6000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 2.010,00 kr
    2.010,00 kr
    1 * 1 ROL
       2.010,00 kr
  • GAINJTTA-242C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    44,5×19,1 mm (1,75×0,75")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.260,00 kr
    1.260,00 kr
    1 * 1 ROL
       1.260,00 kr
  • GAINJTTA-24C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    44,5×25,4 mm (1,75×1")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 937,00 kr
    937,00 kr
    1 * 1 ROL
       937,00 kr
  • GAINJTTA-24C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    44,5×25,4 mm (1,75×1")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 937,00 kr
    937,00 kr
    1 * 1 ROL
       937,00 kr
  • GAINJTTA-37C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    48×31,8 mm (1,89×1,25")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 790,00 kr
    790,00 kr
    1 * 1 ROL
       790,00 kr
  • GAINJTTA-128C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    50,8×19,1 mm (2×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 481,00 kr
    481,00 kr
    1 * 1 ROL
       481,00 kr
  • GAINJTTA-128C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    50,8×19,1 mm (2×0,75")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 961,00 kr
    961,00 kr
    1 * 1 ROL
       961,00 kr
  • GAINJTTA-28C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    50,8×25,4 mm (2×1")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
  • GAINJTTA-28C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    50,8×25,4 mm (2×1")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
  • GAINJTTA-14C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    50,8×38,1 mm (2×1,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 984,00 kr
    984,00 kr
    1 * 1 ROL
       984,00 kr
  • GAINJTTA-14C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    50,8×38,1 mm (2×1,5")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.970,00 kr
    1.970,00 kr
    1 * 1 ROL
       1.970,00 kr
  • GAINJTTA-27C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    50,8×50,8 mm (2×2")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
  • GAINJTTA-27C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    50,8×50,8 mm (2×2")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
  • GAINJTTA60NP10.5WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    50,8×76,2 mm (2×3")
    500 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 641,00 kr
    641,00 kr
    1 * 1 ROL
       641,00 kr
  • GAINJTTA-206C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    54×6,9 mm (2,12×0,27")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 601,00 kr
    601,00 kr
    1 * 1 ROL
       601,00 kr
  • GAINJTTA-231C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    57,2×25,4 mm (2,25×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 800,00 kr
    800,00 kr
    1 * 1 ROL
       800,00 kr
  • GAINJTTA-32C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    63,5×12,7 mm (2,5×0,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 450,00 kr
    450,00 kr
    1 * 1 ROL
       450,00 kr
  • GAINJTTA-32C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    63,5×12,7 mm (2,5×0,5")
    3000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.350,00 kr
    1.350,00 kr
    1 * 1 ROL
       1.350,00 kr
  • GAINJTTA-68C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    63,5×19,1 mm (2,5×0,75")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
  • GAINJTTA-68C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    63,5×19,1 mm (2,5×0,75")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
  • GAINJTTA-5C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    63,5×25,4 mm (2,5×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 840,00 kr
    840,00 kr
    1 * 1 ROL
       840,00 kr
  • GAINJTTA-5C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    63,5×25,4 mm (2,5×1")
    4000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 3.360,00 kr
    3.360,00 kr
    1 * 1 ROL
       3.360,00 kr
  • GAINJTTA-238C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    63,5×38,1 mm (2,5×1,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 824,00 kr
    824,00 kr
    1 * 1 ROL
       824,00 kr
  • GAINJTTA-11C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    67,1×7 mm (2,64×0,28")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 601,00 kr
    601,00 kr
    1 * 1 ROL
       601,00 kr
  • GAINJTTA-181C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    70×38,1 mm (2,75×1,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 903,00 kr
    903,00 kr
    1 * 1 ROL
       903,00 kr
  • GAINJTTA-181C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    70×38,1 mm (2,75×1,5")
    2000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.810,00 kr
    1.810,00 kr
    1 * 1 ROL
       1.810,00 kr
  • GAINJTTA-36C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    76,2×25,4 mm (3×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 682,00 kr
    682,00 kr
    1 * 1 ROL
       682,00 kr
  • GAINJTTA-36C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    76,2×25,4 mm (3×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 682,00 kr
    682,00 kr
    1 * 1 ROL
       682,00 kr
  • GAINJTTA-56C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    76,2×50,8 mm (3×2")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
  • GAINJTTA-56C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    76,2×50,8 mm (3×2")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
  • GAINJTTA99C10.25WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    76,2×76,2 mm (3×3")
    250 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 472,00 kr
    472,00 kr
    1 * 1 ROL
       472,00 kr
  • GAINJTTA-101C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    76×38,1 mm (3×1,5")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 984,00 kr
    984,00 kr
    1 * 1 ROL
       984,00 kr
  • GAINJTTA122C10.3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    101,6×101,6 mm (4×4")
    300 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 987,00 kr
    987,00 kr
    1 * 1 ROL
       987,00 kr
  • GAINJTTA-127C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    101,6×25,4 mm (4×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 896,00 kr
    896,00 kr
    1 * 1 ROL
       896,00 kr
  • GAINJTTA-127C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    hvid
    101,6×25,4 mm (4×1")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 896,00 kr
    896,00 kr
    1 * 1 ROL
       896,00 kr
  • GAINJTTA62C1-0.5WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    101,6×50,8 mm (4×2")
    500 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 847,00 kr
    847,00 kr
    1 * 1 ROL
       847,00 kr
  • GAINJTTA-242C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    hvid
    44,5×19,1 mm (1,75×0,75")
    1000 labels/roll (1 label across)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 419,00 kr
    419,00 kr
    1 * 1 ROL
       419,00 kr