Orders LinkContactUsLinkComponent
Glass cleaners, SURE®
Glass cleaners, SURE®
Glass cleaner, 6×0,75 L bottle
Katalog # 141-1570
   Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage
Leverandør:   Diversey
Glass cleaners, SURE®
Katalog # 141-1570
Leverandør:  Diversey
   Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage
Glass cleaners, SURE®
Glass cleaners, SURE®
Glass cleaner, 6×0,75 L bottle
Price
237,00 kr
Log ind for tilgængelighed
1 * 6 Fl
Product Details & Documents
Ready to use spray cleaner for streak-free glass cleaning.
  • Excellent removal of fingermarks as well as the spot treatment of other slightly adherent soils
  • Easy spray-on, wipe-off action
  • No fragrance nor dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product for user
SURE® glass cleaner is part of a unique range of plant-based, Cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test
EU-Ecolabel (EU: NL/020/045)
Specifikationer
  • Beskrivelse:
    Glass cleaner
  • Seq:
    01
  • Pakket:
    6×0,75 L bottle
  • Cat. No.:
    141-1570
  • Environmentally Preferable:
    Bæredygtige materialer|Sikrere sundhed for mennesker og miljø|Bæredygtig emballage