Mounting medium for histology/microscopy, Histokitt, Assistent®
Catalog # HECH41025010
Supplier: GLASWARENFABRIK KARL HECHT
CAS Number:
Specifications
- Pakn:500 mL
- Beskrivelse:Histokitt mounting medium
- Environmentally Preferable:
Specifications
About this item
Mounting medium for histology/microscopy.
- Light-resistant
- Neutral
- Colourless
- Does not affect colour of the specimen
- Soluble e.g. In benzene, chloroform, dioxane, xylene
- In UN certified tinplate bottle
Hardening time 20 minutes, cold-resistant down to -17 °C (+1,4 °F).