Orders LinkContactUsLinkComponent
Yeast viability staining kits
Yeast viability staining kits
Leverandør:   Biotium
Yeast viability staining kits
Leverandør:  Biotium
Certifikater
Yeast viability staining kits
3 tilgængelige muligheder nedenfor
Product Details & Documents
The Yeast Viability Staining Kit is a fluorescent assay to determine yeast viability. Each kit contains two fluorescent components: CF® Dye-labeled Concanavalin A (Con A) and Live-or-Dye™ viability dye. Biotium offers 3 different color combinations to choose from.
  • Stable and selective dead cell staining
  • Simple, rapid, no-wash staining protocol
  • Tolerates fixation and permeabilization
  • Suitable for flow cytometry and microscopy
  • Three color combinations to choose from
When both dyes are used to stain yeast, live cells display bright fluorescent cell wall staining (Con A), while dead cells with compromised membrane activity exhibit both cell wall staining and cytoplasmic staining (Live-or-Dye™). The dye labeling is extremely stable, allowing the cells to be fixed and permeabilized without loss of fluorescence or dye transfer between cells.
Live-or-Dye™ viability dyes are cell membrane impermeable amine-reactive dyes designed for discrimination between live and dead cells during flow cytometry or microscopy. They are able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins.
Concanavalin A (Con A) is a widely used lectin that selectively binds to α-mannopyranosyl and α-glucopyranosy residues, found in the cell wall of yeast and fungi. It is labeled with one of our superior bright and photostable CF® Dyes.
Green/Red: CF®488A Con A and Live-or-Dye™ 594/614
Red/Blue: CF®594 Con A and Live-or-Dye™ 405/452
Far-Red/Red: CF®640R Con A and Live-or-Dye™ 568/583
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 31063-1
  • 31063-2
  • 31063-3
  • Farve:
  • Green/Red
  • Red/Blue
  • Red/Far-red
Specifikationer
Product Family Options
Produktinformation
  • Pk Info
    Pakn
    Tilgængelighed
    Price
  • 31063-1
    Yeast viability staining kit, Green/Red
    Yeast viability staining kit, Green/Red
    1000 assays
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 5.270,00 kr
    5.270,00 kr
    1 * 1 Each
       5.270,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    31063-1
    Farve
    Green/Red
  • 31063-2
    Yeast viability staining kit, Red/Blue
    Yeast viability staining kit, Red/Blue
    1000 assays
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 5.270,00 kr
    5.270,00 kr
    1 * 1 Each
       5.270,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    31063-2
    Farve
    Red/Blue
  • 31063-3
    Yeast viability staining kit, Red/Far-red
    Yeast viability staining kit, Red/Far-red
    1000 assays
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 5.270,00 kr
    5.270,00 kr
    1 * 1 Each
       5.270,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    31063-3
    Farve
    Red/Far-red