Test strips, QUANTOFIX Chlorine Sensitive 1
QUANTOFIX Chlorine Senstivie 1, 0 • 0.05 • 0.1 • 0.2 • 0.4 • 0.8 • 1.2 - mg/L - Cl₂, Redox reaction, 40×40×100 mm (package), 50 test(s), 50
Leverandør: MACHEREY-NAGEL
Test strips, QUANTOFIX Chlorine Sensitive 1
Katalog # 634-1189
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Semi-quantitative test strips for reliable determination of chlorine concentrations as low as 0,05 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity by a factor of nearly 10. Ideal for rapid and easy analysis of chlorine directly on-site. No maintenance or calibration is required.
- Sensitive rapid test
Documents
Specifikationer
Specifikationer
- Kemisk metode:Redox reaction
- Pakn:50
- Beskrivelse:QUANTOFIX Chlorine Senstivie 1
- Dimensioner:40×40×100 mm (package)
- Test pr. sæt:50 test(s)
- Område:0 • 0.05 • 0.1 • 0.2 • 0.4 • 0.8 • 1.2 - mg/L - Cl₂
- Indhold:50 test strips in a tube, instruction leaflet
- Cat. No.:634-1189
- Overensstemmelse:No hazardous material
- Platform:Semi-quantitative test strips
- Holdbarhed (måneder):30 (from production)
- Vægt:32 g
- Instrument:No
- Parameter:All reagents included
- Method:Chlorine - Redox reaction (white → blue-green)