Orders LinkContactUsLinkComponent
HiTrap columns, Phenyl FF (High Sub)
HiTrap columns, Phenyl FF (High Sub)
Supplier:   Cytiva
HiTrap columns, Phenyl FF (High Sub)
Supplier:  Cytiva
Certificates
HiTrap columns, Phenyl FF (High Sub)
2 Options Available Below
Product Details & Documents
Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.
  • Provide convenient and fast small-scale protein purifications
  • Two levels of substitution on Phenyl Sepahrose 6 Fast Flow help find optimal selectivity and binding capacity
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
HiTrap Phenyl FF (high sub) are prepacked Phenyl Sepharose 6 Fast Flow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.
Specifications for Product Family
Specifications
  • Catalog No:
  • 17-1353-01
  • 17-1355-01
  • Maks. tryk:
  • 5 bar
  • 5 bar
  • pH-område:
  • 3 - 13
  • 3 - 13
Specifications
Product Family Options
Product Information
  • Beskrivelse
    Kolonnemateriale
    Partikelstørrelse
    Sengens dimensioner
    Kapacitet
    Flowhastighed
    Availability
    Price
  • HiTrap™ phenyl FF (low sub)
    PP
    90 µm
    7×25 mm
    1 ml
    <1 ml/min
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 1.870,00 kr
    1.870,00 kr
    1 * 5 Items
       1.870,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    17-1353-01
    Maks. tryk
    5 bar
    pH-område
    3 - 13
  • HiTrap™ phenyl FF (high sub)
    PP
    90 µm
    7×25 mm
    1 ml
    <1 ml/min
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 1.860,00 kr
    1.860,00 kr
    1 * 5 Items
       1.860,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    17-1355-01
    Maks. tryk
    5 bar
    pH-område
    3 - 13