About this item
Whatman Puradisc micron syringe filters from Cytiva's offer a versatile solution for analytical sample preparation needs. They are a variety of options available to suit all labs needs, and are well-suited for routine syringe filtration of samples up to 100 mL for a range of applications, including high performance liquid chromatography (HPLC) and capillary electrophoresis (CE).
- Choice of 13 mm or 25 mm diameter nonsterile syringe filter to suit filtration sample volume
- Sub-micron pore sizes, 0,2 μm and 0,45 μm, for fine particle retention to protect instruments and chromagraphy columns
- Autoclavable for sterile sample preparation
- Also available with depth polypropylene (DpPP) filtration medium
- Recyclable after use
Whatman polypropylene syringe filters have high and uniform tolerance to heat and mechanical stress. The slightly hydrophobic membrane is resistant to a wide variety of organic solvents.
Cytiva's Whatman Puradisc line is a comprehensive syringe filter portfolio used in a variety of filtration applications. Puradisc is available in a range of filtration media to filter samples of diverse chemical solvents, compounds and sample types. Filtration materials include: regenerated cellulose (RC), cellulose nitrate, nylon, polytetrafluoroethylene (PTFE), polyethersulfone (PES), polypropylene, polyvinylidene difluoride (PVDF), and cellulose fiber acetate.
Puradisc syringe filters are well-suited for a wide variety of sample types and applications, including capillary electrophoresis(CE), biological sample preparations and analysis, protein filtration, UV spectroscopy and high performance liquid chromatography (HPLC).
Cytiva also offers high-performance GD/X syringe filters with a multi-layer glass fiber prefilter for filtering larger volumes of harder-to-filter samples.
Specifications
- Catalog No:
- 514-0237
- 514-8030
- 516-1434
- 514-8032
- Ø:
- 25 mm
- 25 mm
- 25 mm
- 13 mm
- Mål:
- 22,9×28,4 mm
- 22,9×28,4 mm
- 22,9×28,4 mm
- 16,3×19,8 mm
- Max. operating pressure:
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- Vægt:
- 2,7 g
- 2,7 g
- 2,7 g
- 0,95 g
Specifications
Related Information
Certificates
Search by Lot Number28420315_INSTRUCTIONS.pdf 28420316_INSTRUCTIONS.pdf 28420320_INSTRUCTIONS.pdf 28420321_INSTRUCTIONS.pdf
Manual
CoC_WHAT6788-2504_17232873 CoC_WHAT6788-2504_17288316 CoC_WHAT6788-2504_18033650 CoC_WHAT6790-2504_17340353 CoC_WHAT6790-2504_17413309
WHAT6788-2502_TDS_EN